Skip to Content
Merck
All Photos(3)

Documents

203076

Sigma-Aldrich

Cobalt

granular, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12141710
eCl@ss:
38190601
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

granular

resistivity

6.24 μΩ-cm, 20°C

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the synthesis of a metal-rich layered telluride, TaCo2Te2, which possesses ideal characteristics for photovoltaic applications, for surface scanning studies, and which may give clean and atomically flat surfaces.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 4 - Carc. 1B - Eye Irrit. 2 - Flam. Sol. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuhasen, J. et al.
Chemistry of Materials, 10, 3870-3870 (1998)
Jeannette Ø Penny et al.
Acta orthopaedica, 84(2), 130-137 (2013-04-20)
Wear particles from metal-on-metal arthroplasties are under suspicion for adverse effects both locally and systemically, and the DePuy ASR Hip Resurfacing System (RHA) has above-average failure rates. We compared lymphocyte counts in RHA and total hip arthroplasty (THA) and investigated
S Perconti et al.
Journal of biological regulators and homeostatic agents, 27(2), 443-454 (2013-07-09)
Size-dependent characteristics of novel engineered nanomaterials might result in unforeseen biological responses and toxicity. To address this issue, we used cDNA microarray analysis (13443 genes) coupled with bioinformatics and functional gene annotation studies to investigate the transcriptional profiles of Balb/3T3
Heather De Bari et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 69(Pt 3), 228-236 (2013-03-23)
The X-ray crystal structure of ribosome hibernation promoting factor (HPF) from Vibrio cholerae is presented at 2.0 Å resolution. The crystal was phased by two-wavelength MAD using cocrystallized cobalt. The asymmetric unit contained two molecules of HPF linked by four Co
Hassan Imran Afridi et al.
Clinical laboratory, 59(3-4), 247-256 (2013-06-04)
The aim of the present study was to compare the level of chromium (Cr), cobalt (Co), and manganese (Mn) in biological samples (blood, urine, and scalp hair) of patients suffering from different types of viral hepatitis (A, B, C, D

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service