Skip to Content
Merck
All Photos(1)

Documents

202207

Sigma-Aldrich

Boron tribromide

ReagentPlus®, 99.9%

Synonym(s):

Tribromoboron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BBr3
CAS Number:
Molecular Weight:
250.52
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.6 (vs air)

vapor pressure

40 mmHg ( 14 °C)

product line

ReagentPlus®

Assay

99.9%

color

colorless

bp

~90 °C (lit.)

mp

−46 °C (lit.)

density

2.60 g/mL at 20 °C (lit.)

SMILES string

BrB(Br)Br

InChI

1S/BBr3/c2-1(3)4

InChI key

ILAHWRKJUDSMFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Drug intermediate 6-nitro-L-DOPA
  • Luminescent polystyrene derivatives with sterically protected carbazolylborane moieties
  • High-quality boron-doped graphene via Wurtz-type reductive coupling reaction
  • Mercapto-(+)-methamphetamine haptens for synthesis of (+)-methamphetamine conjugate vaccines with improved epitope densities
  • Micrometer-sized organic molecule-DNA hybrid structures
  • Borane complexes via electrophilic aromatic borylation reactions
  • A 5-HT2C receptor agonist
  • Biphenyl-derivatives possessing tertiary amino groups as β-secretase(BACE1) inhibitors for the treatment of Alzheimer′s disease
  • A highly near-IR region fluorescent p-extended boron aza-dipyrromethene moiety unit
  • Tetrahydroisoquinoline derivatives via intramolecular cyclization of methoxy-substituted N-phenethylimides
Used to cleave aryl methyl ethers in a syntheses of a benzopyranobenzopyran from a coumarin scaffold and ladder-type conjugated polymers.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 8689-8689 (2006)
Xiu-Yan Dong et al.
Molecules (Basel, Switzerland), 23(5) (2018-04-26)
A novel heterotrinuclear complex [Cu₂(L)Na(µ-NO₃)]∙CH₃OH∙CHCl₃ derived from a symmetric bis(salamo)-type tetraoxime H₄L having a naphthalenediol unit, was prepared and structurally characterized via means of elemental analyses, UV-Vis, FT-IR, fluorescent spectra and single-crystal X-ray diffraction. The heterobimetallic Cu(II)⁻Na(I) complex was acquired
Pengpeng Lu et al.
Carbohydrate polymers, 216, 149-156 (2019-05-03)
A novel porous β-cyclodextrin/pillar[5]arene copolymer was prepared using tetrafluoroterephthalonitrile as cross-linker for rapid removal of variety of organic pollutants. The copolymer was characterized by water contact angle, scanning electron microscope, FT-IR spectrum, solid-state 13C NMR spectrum, thermo-gravimetric analysis, nitrogen adsorption-desorption
Lu-Mei Pu et al.
Molecules (Basel, Switzerland), 23(4) (2018-04-05)
[Co₂(L)Ce(OAc)₃(CH₃CH₂OH)]·1.5CH₃OH∙0.5CH₂Cl₂, a heterotrinuclear Co(II)-Ce(III) bis(salamo)-type complex with a symmetric bi(salamo)-type ligand H₄L and an acyclic naphthalenediol moiety, was designed, synthesized and characterized by elemental analyses, FT-IR, UV-Vis and fluorescence spectroscopy and X-ray crystallography. The X-ray crystallographic investigation revealed the heterotrinuclear
Jing Hao et al.
Materials (Basel, Switzerland), 11(4) (2018-03-30)
A novel, simple, highly selective, and sensitive fluorescence chemosensor for detecting Cd2+ that was constructed from a bis(salamo)-type compound (H₄L) with two N₂O₂ chelating moieties as ionophore was successfully developed. Sensor H₄L could show fluorescence turn-on response rapidly and significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service