Skip to Content
Merck

202118

Hafnium(IV) oxide

powder, 98%

Synonym(s):

Hafnia, Hafnium dioxide, Hafnium oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
HfO2
CAS Number:
Molecular Weight:
210.49
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352303
EC Number:
235-013-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98%

form

powder

composition

HfO₂

reaction suitability

core: hafnium

density

9.68 g/mL at 25 °C (lit.)

SMILES string

O=[Hf]=O

InChI

1S/Hf.2O

InChI key

CJNBYAVZURUTKZ-UHFFFAOYSA-N

Application

  • Hafnium(IV) oxide: Utilized predominantly in the semiconductor industry, Hafnium(IV) oxide offers excellent thermal and chemical stability, which makes it suitable as a high-k gate dielectric material in metal-oxide-semiconductor (MOS) devices. Its application has become increasingly important with the miniaturization of electronic components, aiding in the enhancement of transistor performance without further reducing the component size. This role is critical in the development of more efficient, faster computing technologies (Sigma-Aldrich, CAS 12055-23-1).


Still not finding the right product?

Explore all of our products under Hafnium(IV) oxide


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library