Skip to Content
Merck
All Photos(3)

Key Documents

193291

Sigma-Aldrich

2-Chloropyrimidine

95%

Synonym(s):

2-Chloro-1,3-pyrimidine, Pyrimidin-2-yl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3ClN2
CAS Number:
Molecular Weight:
114.53
Beilstein:
107171
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

crystals

bp

75-76 °C/10 mmHg (lit.)

mp

63-66 °C (lit.)

functional group

chloro

SMILES string

Clc1ncccn1

InChI

1S/C4H3ClN2/c5-4-6-2-1-3-7-4/h1-3H

InChI key

UNCQVRBWJWWJBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloropyrimidine undergoes cobalt-catalyzed cross-coupling reaction with aryl halides.

Application

2-Chloropyrimidine was used in the synthesis of:
  • novel bis(2-(pyrimidin-2-yl)ethoxy)alkanes
  • 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay
  • cis- and trans-octahydropyrrolo[2,3]pyridine derivatives
  • 2-amino-4-heteroarylpyrimidines

Other Notes

Remainder 2-hydroxypyrimidine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew G Bursavich et al.
Organic letters, 7(19), 4113-4116 (2005-09-09)
[reaction: see text] An expedient synthesis of diverse 2-amino-4-heteroarylpyrimidines via a 2-chloropyrimidine intermediate is described. A series of potentially biologically active analogues have been synthesized in two parallel steps to afford focused arrays.
Vangavaragu Jhansi Rani et al.
Archiv der Pharmazie, 345(8), 663-669 (2012-05-18)
The pyrimidine nucleus is an important component of nucleic acids (DNA and RNA) and vitamins (B(2) and folic acid). It is evident from the literature that pyrimidine derivatives possess a wide spectrum of biological activities such as antioxidant, anticancer, antibacterial
Vikas S Padalkar et al.
Chemistry Central journal, 5, 72-72 (2011-11-10)
Fluorescent dyes with biocompatible functional group and good fluorescence behavior are used as biosensor for monitoring different biological processes as well as detection of protein assay. All reported fluorophore used as sensors are having high selectivity and sensitivity but till
Carina Sollert et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(14), 5380-5386 (2015-02-18)
The Ru-catalysed C2-H arylation of indoles and pyrroles by using boronic acids under oxidative conditions is reported. This reaction can be applied to tryptophan derivatives and tolerates a wide range of functional groups on both coupling partners, including bromides and
Igor Goljer et al.
Chirality, 21(7), 681-691 (2008-09-17)
Reaction of (S)- or (R)-3-aminoquinuclidine with 2-chloropyrimidine or 2-bromopyrimidine led to an unexpected formation of both cis- and trans-octahydropyrrolo [2,3]pyridine derivatives. A single-step synthesis of two of the four stereoisomers of these octahydropyrrolo[2,3]pyridine derivatives provides a convenient way of generating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service