Recommended Products
Assay
99%
form
solid
mp
112-113 °C (lit.)
SMILES string
CCc1ccc(cc1)C(O)=O
InChI
1S/C9H10O2/c1-2-7-3-5-8(6-4-7)9(10)11/h3-6H,2H2,1H3,(H,10,11)
InChI key
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
4-Ethylbenzoic acid reacts with lanthanum nitrate in aqueous solution to yield the polymer catena-poly[[aqua(4-ethylbenzoic acid-κO)lanthanum(III)]-tri-μ-4-ethylbenzoato].
Application
4-Ethylbenzoic acid was used in the synthesis of ethyl 4-vinyl-α-cyano-β-phenylcinnamate. It was also used to functionalize the edge of “pristine” graphite in the presence of polyphosphoric acid/phosphorus pentoxide.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Functional Polymers. VII. Ethyl 4-Vinyl-α-cyano-β-phenylcinnamate.
Polymer Journal, 13(6), 521-536 (1981)
Acta crystallographica. Section E, Structure reports online, 66(Pt 2), m183-m184 (2010-01-01)
The reaction of lanthanum nitrate and 4-ethyl-benzoic acid (EBAH) in aqueous solution yielded the title polymer, [La(C(9)H(9)O(2))(3)(C(9)H(10)O(2))(H(2)O)](n). The asymmetric unit contains one La(III) atom, three 4-ethyl-benzoate (EBA) ligands, one neutral EBAH ligand and one coordinated water mol-ecule. Each La(III) ion
Science (New York, N.Y.), 235(4788), 593-596 (1987-01-30)
Increasing quantities of man-made organic chemicals are released each year into the biosphere. Some of these compounds are both toxic and relatively resistant to physical, chemical, or biological degradation, and they thus constitute an environmental burden of considerable magnitude. Genetic
Journal of agricultural and food chemistry, 48(2), 407-412 (2000-02-26)
Changes in flavor release and aroma characteristics in a medium including food phenolics may be attributed to an intermolecular interaction between flavor compounds and phenolics. To investigate the interaction, one- and two-dimensional NMR studies have been carried out on the
Ruthenium(II)-catalyzed synthesis of hydroxylated arenes with ester as an effective directing group.
Organic letters, 14(11), 2874-2877 (2012-05-16)
An unprecedented Ru(II) catalyzed ortho-hydroxylation has been developed for the facile synthesis of a variety of multifunctionalized arenes from easily accessible ethyl benzoates with ester as an efficient directing group. Both the TFA/TFAA cosolvent system and oxidants serve as the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service