Skip to Content
Merck
All Photos(1)

Documents

189162

Sigma-Aldrich

2-Methoxy-1,4-naphthoquinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O3
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

184-187 °C (lit.)

SMILES string

COC1=CC(=O)c2ccccc2C1=O

InChI

1S/C11H8O3/c1-14-10-6-9(12)7-4-2-3-5-8(7)11(10)13/h2-6H,1H3

InChI key

OBGBGHKYJAOXRR-UHFFFAOYSA-N

General description

2-Methoxy-1,4-naphthoquinone is a potential candidate for Helicobacter pylori infection related disease therapy. It is isolated from the leaves of Impatiens glandulifera.

Application

2-Methoxy-1,4-naphthoquinone was used in the preparation of 2-(4-X-phenylene)amine-1,4-naphthoquinones (X= ferrocenyl, OMe, Me, I, Cl and NO2).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General method for the high yield preparation of 2-(4-X-phenylene) amine-1, 4-naphthoquinones (X= ferrocenyl, OMe, Me, I, Cl, and NO2) from 2-methoxy-1, 4-naphthoquinone and investigation of H+ and Mg2+ catalysts with DFT calculations.
Francisco AI, et al.
Journal of Molecular Structure, 891(1), 228-232 (2008)
K Ishiguro et al.
Journal of natural products, 61(9), 1126-1129 (1998-09-28)
Dinaphthofuran-7,12-dione derivatives named balsaminones A (1) and B (2) were isolated from the pericarp of Impatiens balsamina L. together with the known compound 2-methoxy-1,4-naphthoquinone (3). Their structures were elucidated by spectral techniques. These compounds have significant antipruritic activity.
Scott B Vafai et al.
PloS one, 11(9), e0162686-e0162686 (2016-09-14)
Deficiency of mitochondrial complex I is encountered in both rare and common diseases, but we have limited therapeutic options to treat this lesion to the oxidative phosphorylation system (OXPHOS). Idebenone and menadione are redox-active molecules capable of rescuing OXPHOS activity
Aldana L Zalazar et al.
Food research international (Ottawa, Ont.), 116, 916-924 (2019-02-06)
Probabilistic microbial modelling using logistic regression was used to predict the growth/no growth (G/NG) interfaces of Zygosaccharomyces bailii in simulated acid sauces as a function of natamycin, xanthan gum (XG) and sodium chloride concentrations. The growth was assessed colorimetrically by
Naomi Mori et al.
Journal of natural medicines, 65(1), 234-236 (2010-10-05)
A screening study using a luciferase assay to identify natural products which inhibit Wnt signaling was carried out. The bioassay-guided fractionation of aerial parts of a plant, Impatiens balsamina, led to the isolation of 2-methoxy-1,4-naphthoquinone (1) as an active compound.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service