Skip to Content
Merck
All Photos(1)

Documents

186783

Sigma-Aldrich

2,6-Difluorobenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CN
CAS Number:
Molecular Weight:
139.10
Beilstein:
2045292
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.4875 (lit.)

bp

197-198 °C

mp

25-28 °C (lit.)

density

1.246 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(F)c1C#N

InChI

1S/C7H3F2N/c8-6-2-1-3-7(9)5(6)4-10/h1-3H

InChI key

BNBRIFIJRKJGEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Difluorobenzonitrile was used in the synthesis of:
  • poly(cyanoaryl ethers) via silyl-method
  • 2-dimethylamino-6-fluorobenzamide
  • phenolphthalein-modified polyarylene ether nitrile copolymers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New polymer syntheses, 15 Syntheses of aromatic polyethers from difluorobenzonitriles and silylated diphenols.
Kricheldorf HR, et al.
Makromol. Chem., Rapid Commun., 8(11), 529-534 (1987)
Xiaocan Liu et al.
Polymers, 11(9) (2019-08-31)
Porous materials with high specific surface area possess a broad application prospect in the treatment of wastewater. In this work, sulfonated poly(arylene ether nitrile) (SPEN) functionalized with a carboxylic acid group was successfully synthesized, which was subsequently transformed into SPEN
Synthesis and laboratory evaluation of 1-(2,6-disubstituted benzoyl)-3-phenylureas, a new class of insecticides. II. Influence of the acyl moiety on insecticidal activity.
K Wellinga et al.
Journal of agricultural and food chemistry, 21(6), 993-998 (1973-11-01)
Synthesis and properties of phenolphthalein-based polyarylene ether nitrile copolymers.
Li C, et al.
Materials Letters, 60(1), 137-141 (2006)
Chenchen Liu et al.
Polymers, 11(7) (2019-07-07)
Poly(arylene ether nitrile)s with sulfonic and carboxylic groups (SCPEN) were synthesized to investigate their electrical properties. This new series of copolymers were prepared by copolymerization of phenolphthalein, potassium hydroquinonesulfonate, and 2,6-difluorobenzonitrile, in different mole ratios. Their thermal, mechanical and dielectric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service