Skip to Content
Merck
All Photos(1)

Key Documents

18265

Sigma-Aldrich

(3-Bromopropyl)trimethoxysilane

≥97.0%

Synonym(s):

γ-Bromopropyltrimethoxysilane, 1-Bromo-3-trimethoxysilylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15BrO3Si
CAS Number:
Molecular Weight:
243.17
Beilstein:
4952577
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.440 (lit.)

bp

130 °C/45 mmHg (lit.)

density

1.298 g/mL at 20 °C (lit.)

functional group

bromo

SMILES string

CO[Si](CCCBr)(OC)OC

InChI

1S/C6H15BrO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-6H2,1-3H3

InChI key

GLISZRPOUBOZDL-UHFFFAOYSA-N

General description

3-bromopropyl trimethoxysilane is a bromo silane. It is popularly used as a silane coupling agent.

Application

3-bromopropyl trimethoxysilane (BPTS) acted as initiating sites for functionalization magnetite (Fe3O4) nanoparticles.{31} A study reports the role of BPTS in attaching a polyelectrolyte to an inorganic alumina substrate during the fabrication of a polymeric humidity sensor.{32}

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qiang Li et al.
Journal of colloid and interface science, 511, 285-295 (2017-10-17)
In this work, N-alkylated poly (4-vinylpyridine) (NPVP), a cationic polymer, was firstly applied for the surface modification of Fe
Polymeric humidity sensor using polyelectrolyte derived from poly (amide-sulfone) s.
Jeon YM, et al.
Macromolecular Research, 17(4), 227-231 (2009)
Poly (n-isopropylacrylamide)-based hydrogel coatings on magnetite nanoparticles via atom transfer radical polymerization.
Frimpong RA, et al.
Nanotechnology, 19(17), 175101-175101 (2008)
Servet Tural et al.
Ecotoxicology and environmental safety, 162, 245-252 (2018-07-11)
Click chemistry refers to a group of reactions that are fast, simple to use, easy to purify, versatile, regiospecific, and give high product yields. Therefore, a novel, efficient magnetic nano-sorbent based on N-methyl-D-glucamine attached to magnetic nanoparticles was prepared using
Subodh Kumar et al.
Dalton transactions (Cambridge, England : 2003), 44(26), 11860-11866 (2015-06-10)
Chitosan coated magnetic nanoparticles were synthesized and used as a support for the immobilization of the cobalt(II) acetylacetonate complex [Co(acac)2] and quaternary triphenylphosphonium bromide [P(+)Ph3Br(-)] targeting -NH2 and -OH moieties located on the surface of chitosan. The synthesized material was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service