Skip to Content
Merck
All Photos(3)

Key Documents

181625

Sigma-Aldrich

Poly(Bisphenol A carbonate)

average Mw ~45,000 by GPC

Synonym(s):

4,4′-Dihydroxydiphenyl-2,2-propane carbonate polymer, Bisphenol A carbonate homopolymer, Carbonic acid-4,4′-dihydroxydiphenyl-2,2-propane copolymer, Poly(dian carbonate), Poly[2,2-bis(4-hydroxyphenyl)propane carbonate]

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw ~45,000 by GPC

refractive index

n20/D 1.585

transition temp

Tg 150 °C
Tm 267 °C

density

1.2 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2.CH2O3/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;2-1(3)4/h3-10,16-17H,1-2H3;(H2,2,3,4)

InChI key

XSXWYGABGYBZRM-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

Poly(Bisphenol A carbonate) (PC) is a thermoplastic polymer with optical clarity, high refractive index, and toughness. It can be used to prepare optical lenses and as a supporting matrix to fabricate hybrid nanofiber membranes.

Application

Poly(Bisphenol A carbonate) has been used:
  • To prepare transparent nanocomposites for optical applications.
  • To study the wettability of graphene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Utz et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 128(2), 217-227 (1997-11-14)
A method is proposed for the quantitative measurement of orientational anisotropy in glassy solids based on 2D dipolar NMR spectra with sample flipping (dipolar DECODER experiment). Purely dipolar spectra are obtained by chemical shift refocusing by a multiple pulse sequence.
M Yalon et al.
Journal of cataract and refractive surgery, 14(4), 393-395 (1988-07-01)
Bisphenol-A polycarbonate has been investigated as an improved polymer for ocular implants, especially for intraocular lenses (IOLs). Polycarbonate properties afford special opportunities for development of tougher, stronger, one-piece IOLs. Autoclave or gamma sterilizability and a higher refractive index may provide
L Feenstra et al.
Clinical otolaryngology and allied sciences, 9(4), 215-220 (1984-08-01)
A review is given of the development of the concept of an artificial tympanic membrane. Starting with homologous tympanic membranes we compared biodegradable collagen materials (homologous and heterologous) and biodegradable synthetic materials, poly-glycolic acid, poly-lactic acid and poly-alpha-amino acids. As
Hiroshi Yabu et al.
Langmuir : the ACS journal of surfaces and colloids, 22(11), 4992-4997 (2006-05-17)
We describe here a preparation of pincushion structures with holes, hexagonally arranged microrings, and microdots by simple heating and peeling of self-organized honeycomb-patterned films. We have reported that the honeycomb-patterned films can be prepared by casting the solution of an
Molla R Islam et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(22), 6098-6108 (2011-04-19)
Spontaneous phase-separated, controlled aggregate structures of photo- and electroactive molecules in polymer matrices are of interest for device fabrication. We show that the self-assembly of octabutoxyphthalocyanine (Pc) in polymer matrices leads to tubular morphology of Pc when the film is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service