Skip to Content
Merck
All Photos(1)

Key Documents

180769

Sigma-Aldrich

Ethyl 4-chloroacetoacetate

95%

Synonym(s):

4-Chloro acetoethylacetate, Ethyl 4-chloro-3-oxobutanoate, Ethyl chloroacetoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COCH2CO2C2H5
CAS Number:
Molecular Weight:
164.59
Beilstein:
1761275
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

115 °C/14 mmHg (lit.)

density

1.218 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CCOC(=O)CC(=O)CCl

InChI

1S/C6H9ClO3/c1-2-10-6(9)3-5(8)4-7/h2-4H2,1H3

InChI key

OHLRLMWUFVDREV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 4-chloroacetoacetate was used in the synthesis of phosphorus ylides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Kataoka et al.
Applied microbiology and biotechnology, 51(4), 486-490 (1999-05-26)
The asymmetric reduction of ethyl 4-chloro-3-oxobutanoate (COBE) to ethyl (R)-4-chloro-3-hydroxybutanoate [(R)-CHBE] using Escherichia coli cells, which coexpress both the aldehyde reductase gene from Sporobolomyces salmonicolor and the glucose dehydrogenase (GDH) gene from Bacillus megaterium as a catalyst was investigated. In
Li-Juan Wang et al.
Bioresource technology, 102(14), 7023-7028 (2011-05-17)
An NADH-dependent reductase (ScCR) from Streptomyces coelicolor was discovered by genome mining for carbonyl reductases. ScCR was overexpressed in Escherichia coli BL21, purified to homogeneity and its catalytic properties were studied. This enzyme catalyzed the asymmetric reduction of a broad
K Kita et al.
Applied and environmental microbiology, 65(12), 5207-5211 (1999-12-03)
We cloned and sequenced the gene encoding an NADPH-dependent aldehyde reductase (ARII) in Sporobolomyces salmonicolor AKU4429, which reduces ethyl 4-chloro-3-oxobutanoate (4-COBE) to ethyl (S)-4-chloro-3-hydroxybutanoate. The ARII gene is 1,032 bp long, is interrupted by four introns, and encodes a 37,315-Da
Jer-Yiing Houng et al.
Biotechnology letters, 25(1), 17-21 (2003-07-29)
Amberlite XAD 2 resin enhanced the asymmetric reduction of ethyl 4-chloroacetoacetate (ECA) to S-4-chloro-3-hydroxybutyric acid ethyl ester as catalyzed by Saccharomyces cerevisiae. The absorbed ECA was released slowly to the solution during the reaction so that the substrate inhibition and
I Pieper et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 36(1), 157-170 (2008-12-02)
Computationally predicting the metabolic fates of drugs is a very complex task which is owed not only to the huge and diverse biochemical network in the living cell, but also to the majority of in vivo transformations that occur through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service