Skip to Content
Merck
All Photos(3)

Key Documents

176591

Sigma-Aldrich

Anthracene-d10

98 atom % D

Synonym(s):

1,2,3,4,5,6,7,8,9,10-Decadeuterioanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D10
CAS Number:
Molecular Weight:
188.29
Beilstein:
2055675
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

form

solid

bp

340 °C (lit.)

mp

210-215 °C (lit.)

application(s)

electronics

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c2c([2H])c3c([2H])c([2H])c([2H])c([2H])c3c([2H])c2c1[2H]

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

MWPLVEDNUUSJAV-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzu-Ling Chen et al.
Journal of chromatography. A, 1422, 340-344 (2015-11-01)
The rapid screening of trace amounts of the nitrogenous disinfection by-products, haloacetamides (HAcAms), in drinking and swimming pool water was performed by a simple and reliable procedure based on salt-assisted liquid-liquid extraction (SALLE) combined with injection-port silylation gas chromatography-mass spectrometry
Fairda Malem et al.
Environmental monitoring and assessment, 191(10), 627-627 (2019-09-11)
In this work, we investigated possible contamination of phthalates in seawater and sediment around the eastern coast of Thailand in the area of Chonburi, Rayong, and Chanthaburi. The main focus was on Pradu Bay east of Map Ta Phut, a
Wan-Ting Chang et al.
Marine pollution bulletin, 97(1-2), 217-223 (2015-06-16)
This work investigates the dependence of the sampling rate (Rs) of semi-permeable membrane devices (SPMDs) on flow rate and temperature. The in situ Rs values were obtained using performance reference compounds (PRCs) with weighted polynomial regression and used to estimate
Inyang O Oyo-Ita et al.
Archives of environmental contamination and toxicology, 70(2), 372-382 (2015-11-08)
The distribution of aliphatic and aromatic hydrocarbons in surface sediments of the lower course of the Imo River (Nigeria) was investigated to determine the sources and fate of these compounds. The aliphatic fraction is characterized by a widespread contribution of
Ying Li et al.
Environmental science and pollution research international, 22(23), 18446-18455 (2015-08-14)
The effects of ultrasonic irradiation, the chelating agent modified Fenton reaction, and a combination of ultrasound and the Fenton method in removing petroleum contaminants from a soil were studied. The results showed that the contaminant removal rate of the Fenton

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service