Skip to Content

175676

Ammonium-d4 chloride

98 atom % D

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
ND4Cl
CAS Number:
Molecular Weight:
57.52
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12142200
EC Number:
234-607-9
MDL number:
Isotopic purity:
98 atom % D
Mass shift:
M+4
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1 mmHg ( 160.4 °C)

Quality Segment

isotopic purity

98 atom % D

form

powder

technique(s)

NMR: suitable, bio NMR: suitable

mp

340 °C (subl.) (lit.)

mass shift

M+4

SMILES string

Cl[2H].[2H]N([2H])[2H]

InChI

1S/ClH.H3N/h1H;1H3/i/hD4

InChI key

NLXLAEXVIDQMFP-JBISRTOLSA-N



pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library