Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H11NOS
CAS Number:
Molecular Weight:
241.31
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
229-626-4
MDL number:
Assay:
95%
Form:
powder
Quality Segment
assay
95%
form
powder
mp
180-185 °C (lit.)
functional group
ketone, thioether
SMILES string
CC(=O)c1ccc2Sc3ccccc3Nc2c1
InChI
1S/C14H11NOS/c1-9(16)10-6-7-14-12(8-10)15-11-4-2-3-5-13(11)17-14/h2-8,15H,1H3
InChI key
JWGBOHJGWOPYCL-UHFFFAOYSA-N
Application
2-Acetylphenothiazine was used as a NADPH oxidase (NOX) inhibitor in human platelet functional responses and intracellular signaling pathways. It was also used in the synthesis of 2-phenothiazin-2′-yl-cinchoninic acid derivatives.
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.