Skip to Content
Merck
All Photos(2)

Key Documents

169919

Sigma-Aldrich

Isobutyl methacrylate

97%

Synonym(s):

2-Methylpropyl methacrylate, Isobutyl α-methylacrylate, Isobutyl 2-methyl-2-propenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
142.20
Beilstein:
1747595
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.82 (vs air)

vapor pressure

3.5 mmHg ( 20 °C)

Assay

97%

form

liquid

contains

≤15 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.420 (lit.)

bp

155 °C (lit.)

density

0.886 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)COC(=O)C(C)=C

InChI

1S/C8H14O2/c1-6(2)5-10-8(9)7(3)4/h6H,3,5H2,1-2,4H3

InChI key

RUMACXVDVNRZJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

108.5 °F - closed cup

Flash Point(C)

42.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ricardo Dante Lardone et al.
Journal of biomedical science, 26(1), 67-67 (2019-09-08)
Different neurological disorders frequently display antibodies against several self-glycans. Increasing evidence supports their pathogenic role; however, far less is known about their origin. Meanwhile, antibodies recognizing non-self glycans appear in normal human serum during immune response to bacteria. Using high
Miku Kasai et al.
Sensors (Basel, Switzerland), 20(9) (2020-05-08)
A temperature-sensitive paint (TSP) using a chameleon luminophore [ Tb 0 . 99 Eu 0 . 01 ( hfa ) 3 ( dpbp ) ] n is proposed. The chameleon luminophore was dispersed in isobutyl methacrylate polymer in a toluene
M Caccia et al.
Nature, 562(7727), 406-409 (2018-10-20)
The efficiency of generating electricity from heat using concentrated solar power plants (which use mirrors or lenses to concentrate sunlight in order to drive heat engines, usually involving turbines) may be appreciably increased by operating with higher turbine inlet temperatures
D Tantbirojn et al.
European journal of oral sciences, 108(6), 564-568 (2001-01-12)
An amphiphilic coating is configured as a substantive film that has a tendency for an in-plane two-dimensional polymerization. This coating is hypothesized to protect enamel from in vitro acid decalcification, assessed through the following artificial caries model. Three regions on
Janaina Habib Jorge et al.
Quintessence international (Berlin, Germany : 1985), 40(10), e93-100 (2010-02-18)
To evaluate the effect of microwave postpolymerization heat treatment and water storage time on the cytotoxicity of denture base and reline acrylic resins. Sample disks of 6 acrylic resins were fabricated under aseptic conditions (Kooliner, GC; Tokuyama Rebase II, Tokuyama

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service