Skip to Content
Merck
All Photos(2)

Key Documents

168548

Sigma-Aldrich

1,3-Dichloroacetone

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COCH2Cl
CAS Number:
Molecular Weight:
126.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.38 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥95%

form

solid

bp

173 °C (lit.)

mp

39-41 °C (lit.)

solubility

alcohol: very soluble
diethyl ether: very soluble
water: soluble

density

1.383 g/mL at 25 °C (lit.)

functional group

chloro
ketone

storage temp.

2-8°C

SMILES string

ClCC(=O)CCl

InChI

1S/C3H4Cl2O/c4-1-3(6)2-5/h1-2H2

InChI key

SUNMBRGCANLOEG-UHFFFAOYSA-N

General description

1,3-Dichloroacetone has been identified as a metabolite of 1,3-dichloropropanol by TLC. Kinetics of the reversible hydration of 1,3-dichloroacetone have been studied spectrophotometrically in solutions of water in dioxan and in acetonitrile.

1,3-dichloroacetone is a dihaloketone derivative used as a reagent in the synthesis of complex multicyclic peptides.

Application

1,3-Dichloroacetone was used as acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Muta. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

1, 3-Dichloroacetone
Vittorio and Pace
Synlett, 2010, 2825-2826 (2010)
Han Siean Lee et al.
Biomedicines, 8(10) (2020-10-18)
Relaxin-3 is a highly conserved two-chain neuropeptide that acts through its endogenous receptor the Relaxin Family Peptide-3 (RXFP3) receptor. The ligand/receptor system is known to modulate several physiological processes, with changes in food intake and anxiety-levels the most well studied
Kinetics of the reversible hydration of 1, 3-dichloroacetone in dioxan and acetonitrile solution.
Bell RP, et al.
Proc. Royal Soc. Lond. B., 303(1472), 1-16 (1968)
Xiaoxiao Zhang et al.
Chemosphere, 242, 125227-125227 (2019-11-11)
It is crucial to explore the source, formation process and interdependence of disinfection byproducts (DBPs) to reduce their risk on public health. In this investigation, a source water was chlorinated to evaluate the initial formation rates and the maximum yields
Ling Lu et al.
The Journal of organic chemistry, 64(3), 843-853 (2001-10-25)
Cross-aldol reactions of carbonyl compounds were achieved by the catalysis of SmI(2) or SmI(3), together with molecular sieves, at ambient temperature. 1,3-Dichloroacetone and 1-chloroacetone can be used as acceptor substrates in the cross-aldol reactions with donor substrates such as acetone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service