All Photos(1)
About This Item
Linear Formula:
ClC6H4F
CAS Number:
Molecular Weight:
130.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
form
liquid
refractive index
n20/D 1.501 (lit.)
bp
137-138 °C (lit.)
mp
−43-−42 °C (lit.)
density
1.244 g/mL at 25 °C (lit.)
functional group
chloro
fluoro
SMILES string
Fc1ccccc1Cl
InChI
1S/C6H4ClF/c7-5-3-1-2-4-6(5)8/h1-4H
InChI key
ZCJAYDKWZAWMPR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Chloro-2-fluorobenzene was used in the 9F NMR analysis of conversion of 3,4-fluoro- and 3-chloro-4-fluoroaniline by Rhodococcus sp. strain. It was used in microwave-assisted traceless rapid synthesis of benzimidazolones on Ameba resin.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Haifeng Chen et al.
Chemosphere, 146, 32-39 (2015-12-29)
Halonitromethanes (HNMs), as an emerging class of disinfection by-products containing nitrogen (N-DBPs) in drinking water, have possessed public health concerns. Two most studied materials, graphene and nanometer-sized zero-valent iron, have been successfully combined into binary nanocomposites (G-nZVI) via facile carbonization
Vasili Travkin et al.
FEMS microbiology letters, 209(2), 307-312 (2002-05-15)
In this paper we report the isolation and characterization of an anaerobic enrichment culture as well as of a Rhodococcus sp. strain 2 capable of degrading 3,4-dihaloanilines under nitrate reducing conditions. Using mass spectrometry several of the intermediates formed in
Microwave-assisted traceless synthesis of benzimidazolones.
Xu X-J and Zong Y-X.
Tetrahedron Letters, 48(1), 129-132 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service