Skip to Content
Merck
All Photos(3)

Key Documents

159247

Sigma-Aldrich

12-Aminododecanoic acid

95%

Synonym(s):

12-Aminolauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)11COOH
CAS Number:
Molecular Weight:
215.33
Beilstein:
907502
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

185-187 °C (lit.)

application(s)

peptide synthesis

SMILES string

NCCCCCCCCCCCC(O)=O

InChI

1S/C12H25NO2/c13-11-9-7-5-3-1-2-4-6-8-10-12(14)15/h1-11,13H2,(H,14,15)

InChI key

PBLZLIFKVPJDCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

536.0 °F - closed cup

Flash Point(C)

280 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yasuhisa Asano et al.
Bioscience, biotechnology, and biochemistry, 72(8), 2141-2150 (2008-08-08)
Several omega-laurolactam degrading microorganisms were isolated from soil samples. These strains were capable of growing in a medium containing omega-laurolactam as sole source of carbon and nitrogen. Among them, five strains (T7, T31, U124, U224, and U238) were identified as
K Kamio et al.
Journal of lipid research, 33(8), 1227-1232 (1992-08-01)
For preparation of an affinity ligand, an N-fatty acyl moiety of galactosylceramide (GalCer) was chemically replaced with omega-amino-fatty acid including amino-n-hexanoic acid or amino-n-dodecanoic acid to obtain omega-aminoGalCer. For the synthesis of the compound, galactosylsphingosine (GalSph) was coupled with N-trifluoroacetyl
Hiroto Hayashi et al.
International journal of molecular sciences, 12(9), 5490-5507 (2011-10-22)
Novel poly(ester-urethane)s were prepared by a synthetic route using a lipase that avoids the use of hazardous diisocyanate. The urethane linkage was formed by the reaction of phenyl carbonate with amino acids and amino alcohols that produced urethane-containing diacids and
Mohamed H M Hussein et al.
Journal of surfactants and detergents, 16(2), 233-242 (2013-02-20)
Chitosan is one of the important biopolymers and it is extracted from exoskeletons of crustaceans in sea food waste. It is a suitable eco-friendly carbon steel corrosion inhibitor in acid media; the deacetylation degree of prepared chitosan is more than
Liang Chen et al.
Nanotechnology, 22(10), 105708-105708 (2011-02-04)
The objective of this study is to evaluate the effect of hydroxyapatite (HAP) nanoparticles with different surface charges on the cellular uptake behavior and in vitro cell viability and proliferation of MC3T3-E1 cell lines (osteoblast). The nanoparticles' surface charge was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service