Skip to Content
Merck
All Photos(1)

Key Documents

147826

Sigma-Aldrich

5,6-Dimethoxy-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21
Beilstein:
1241095
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

118-120 °C (lit.)

functional group

ketone

SMILES string

COc1cc2CCC(=O)c2cc1OC

InChI

1S/C11H12O3/c1-13-10-5-7-3-4-9(12)8(7)6-11(10)14-2/h5-6H,3-4H2,1-2H3

InChI key

IHMQOBPGHZFGLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiosemicarbazone derived from 5,6-dimethoxy-1-indanone inhibits bovine viral diarrhea virus infection[1].

Application

5,6-Dimethoxy-1-indanone was used in the synthesis of 2,3-dimethoxy-11H-indeno[1,2-b]quinoline-6,10-dicarboxylic acid[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L W Deady et al.
Bioorganic & medicinal chemistry, 7(12), 2801-2809 (2000-02-05)
A series of ring-substituted analogues of the topoisomerase inhibitor 11-oxo-11H-indeno[1,2-b]quinoline-6-carboxamides was prepared and evaluated. The compounds were prepared by Pfitzinger reaction of the appropriate isatin-7-carboxylic acids and 1-indanones, followed by selective thermal decarboxylation of the resulting tetracyclic diacids, subsequent oxidation
Eliana F Castro et al.
Journal of virology, 85(11), 5436-5445 (2011-03-25)
In the present work, we described the activity of the thiosemicarbazone derived from 5,6-dimethoxy-1-indanone (TSC), which we previously characterized as a new compound that inhibits bovine viral diarrhea virus (BVDV) infection. We showed that TSC acts at a point of
María C Soraires Santacruz et al.
Bioorganic & medicinal chemistry, 25(15), 4055-4063 (2017-06-11)
A series of N
Daniel I Perez et al.
Bioorganic & medicinal chemistry, 17(19), 6914-6925 (2009-09-15)
Thienylhalomethylketones, whose chemical, biological, and pharmaceutical data are here reported, are the first irreversible inhibitors of GSK-3beta described to date. Their inhibitory activity is likely related to the cysteine residue present in the ATP-binding site, which is proposed as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service