Skip to Content
Merck
All Photos(1)

Key Documents

146110

Sigma-Aldrich

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride

99%

Synonym(s):

2,4-Dichlorophenol-6-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OH)SO2Cl
CAS Number:
Molecular Weight:
261.51
Beilstein:
2940993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

80-83 °C (lit.)

SMILES string

Oc1c(Cl)cc(Cl)cc1S(Cl)(=O)=O

InChI

1S/C6H3Cl3O3S/c7-3-1-4(8)6(10)5(2-3)13(9,11)12/h1-2,10H

InChI key

KXFQRJNVGBIDHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride was used in the synthesis of (1R,2R)-(+)-1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)-1,2-diphenylethane and (1R,2R)-(+)- 1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5′-nucleotidase.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective addition of diethylzinc to benzaldehyde catalyzed by chiral titanate complexes with helical ligands.
Guo C, et al.
Tetrahedron, 53(12), 4145-4158 (1997)
A Bertrand et al.
Clinica chimica acta; international journal of clinical chemistry, 119(3), 275-284 (1982-03-12)
We describe a one-step kinetic method for the determination of 5'-nucleotidase (EC 3.1.3.5). Inosine is formed by the hydrolysis of inosine 5'-monophosphate which is catalyzed by seric 5'-nucleotidase, and then is converted to hypoxanthine by nucleoside phosphorylase. Two moles of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service