Skip to Content
Merck
All Photos(2)

Documents

135364

Sigma-Aldrich

Diethyl oxalate

≥99%

Synonym(s):

Diethyl ethanedioate, Ethyl oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCOOC2H5
CAS Number:
Molecular Weight:
146.14
Beilstein:
606350
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.03 (vs air)

Quality Level

vapor pressure

1 mmHg ( 47 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.410 (lit.)

bp

185 °C (lit.)

mp

−41 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
organic solvents: miscible
water: partially soluble

density

1.076 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(=O)OCC

InChI

1S/C6H10O4/c1-3-9-5(7)6(8)10-4-2/h3-4H2,1-2H3

InChI key

WYACBZDAHNBPPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl oxalate undergoes transesterification with phenol in the liquid phase over very efficient MoO3/TiO2 solid-acid sol-gel catalysts to form diphenyl oxalate. It undergoes Claisen condensation with active methylene group of ketosteroids to form glyoxalyl derivatives. It undergoes hydrogenation in the presence of high copper contented mesoporous Cu/SBA-15 catalysts to yield ethylene glycol.

Application

Diethyl oxalate was used in microemulsion synthesis of ZnO nanoparticles. It was also used in the synthesis of sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W W Monafo et al.
Surgery, 80(4), 465-473 (1976-10-01)
The wounds of 60 burned patients were treated topically with cerium nitrate, which was applied either as a cream or in aqueous solution. Cerium nitrate has a potent antiseptic effect in human burn wounds, especially against gram negative bacteria and
Diethyl oxalate as new reagent for spectrophotometric determination of ketosteroids.
S Görög
Analytical chemistry, 42(6), 560-563 (1970-05-01)
Comparison of two novel solution-based routes for the synthesis of equiaxed ZnO nanoparticles.
Elen K, et al.
Journal of Nanomaterials, 2011, 18-18 (2011)
Novel macromolecules containing sym-tetrazine rings and their subsequent reactions through click modifications.
Sayed RA and Wiggins JS.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 49(1), 188-188 (2008)
One pot synthesis of ultra-high copper contented Cu/SBA-15 material as excellent catalyst in the hydrogenation of dimethyl oxalate to ethylene glycol.
Guo X, et al.
Catalysis Letters, 132(1-2), 22-27 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service