Skip to Content
Merck
All Photos(1)

Key Documents

131261

Sigma-Aldrich

Nonanenitrile

98%

Synonym(s):

Octyl cyanide, Pelargononitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CN
CAS Number:
Molecular Weight:
139.24
Beilstein:
1746339
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.6 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.426 (lit.)

bp

224 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

CCCCCCCCC#N

InChI

1S/C9H17N/c1-2-3-4-5-6-7-8-9-10/h2-8H2,1H3

InChI key

PLZZPPHAMDJOSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nonanenitrile was used as an organic nitrogen standard in the determination of a quantitative method for the speciation of ON within ambient atmospheric aerosol[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mustafa Z Özel et al.
Environmental science & technology, 45(4), 1497-1505 (2011-01-08)
Atmospheric aerosols contain a highly complex mixture of organic and inorganic compounds; however, as a chemical class relatively little is known about organic nitrogen (ON) content, with few satisfactory methods for speciated analysis. In this paper we report a sensitive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service