Skip to Content
Merck
All Photos(1)

Documents

130966

Sigma-Aldrich

α-Chlorophenylacetyl chloride

technical grade, 90%

Synonym(s):

α-Chloro-α-phenylacetyl chloride, α-Chlorobenzeneacetyl chloride, (±)-2-Chloro-2-phenylacetyl chloride, 2-Chloro-2-phenylacetyl chloride, 2-Phenyl-2-chloroacetyl chloride, Phenyl(chloro)acetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHClCOCl
CAS Number:
Molecular Weight:
189.04
Beilstein:
1073019
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.544 (lit.)

bp

120 °C/23 mmHg (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

ClC(C(Cl)=O)c1ccccc1

InChI

1S/C8H6Cl2O/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H

InChI key

FGEAOSXMQZWHIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-Chlorophenylacetyl chloride was used in the synthesis of α-chlorophenylacetic acid (CPAA). It was also used to prepare chloro-telechelic macroinitiators.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3 - Water-react 3

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

123.8 °F

Flash Point(C)

51 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

POLY (VINYLBENZYLPHOSPHONIC ACID)-BASED PROTON CONDUCTORS AS POLYMER ELECTROLYTE MEMBRANES FOR FUEL CELLS APPLICATIONS.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 49(1), 556-556 (2008)
Consecutive living polymerization from cationic to radical: a straightforward yet versatile methodology for the precision synthesis of ?cleavable? block copolymers with a hemiacetal ester junction.
Ouchi M, et al. et al.
Polym. Chem., 3, 2193-2199 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service