Skip to Content
Merck
All Photos(2)

Key Documents

126993

Sigma-Aldrich

N-Ethylbenzylamine

97%

Synonym(s):

N-Benzylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHCH2CH3
CAS Number:
Molecular Weight:
135.21
Beilstein:
386023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

191-194 °C (lit.)

density

0.909 g/mL at 25 °C (lit.)

functional group

amine
phenyl

SMILES string

CCNCc1ccccc1

InChI

1S/C9H13N/c1-2-10-8-9-6-4-3-5-7-9/h3-7,10H,2,8H2,1H3

InChI key

HVAAHUDGWQAAOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

N-Ethylbenzylamine has been used to demonstrate the reactivity of NDTE (2,5-dihydroxyphenylacetic acid, 2,5-bis-tetrahydropyranyl ether p-nitrophenyl ester) and HLTE (homogentisic gamma-lactone tetrahydropyranyl ether). It has also been used to prepare 2-[(N-Benzyl, N-ethyl)amino]derivative using 2-chloro-4H-pyrido[1,2-a]pyrimidin-4-one.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M J Rose et al.
Analytical chemistry, 71(11), 2221-2230 (1999-06-15)
Two new reagents, NDTE (2,5-dihydroxyphenylacetic acid, 2,5-bis-tetrahydropyranyl ether p-nitrophenyl ester) and HLTE (homogentisic gamma-lactone tetrahydropyranyl ether), are described for the chemical derivatization of primary and/or secondary amines to form an electrochemically active product. These reagents undergo reaction with the aforementioned
M Di Braccio et al.
Il Farmaco; edizione scientifica, 43(9), 705-723 (1988-09-01)
The N,N-disubstituted 4-amino-2H-pyrido[1,2-a]pyrimidin-2-ones (III) and isomer 2-amino-4H-pyrido[1,2-a]pyrimidin-4-ones (IV) were obtained from the reaction of 2-aminopyridine with the N,N-disubstituted ethyl malonamate/phosphorus oxychloride reagent (II), in refluxing 1,2-dichloroethane. 2-[(N-Benzyl, N-ethyl)amino]derivative (IV b) was also prepared in excellent yield by treating 2-chloro-4H-pyrido[1,2-a]pyrimidin-4-one (V)
Md Salman Shakil et al.
Biomedicines, 9(2) (2021-01-31)
Hydroxypyr(id)ones are a pharmaceutically important class of compounds that have shown potential in diverse areas of drug discovery. We investigated the 3-hydroxy-4-pyridones 1a-1c and 3-hydroxy-4-thiopyridones 1d-1f as well as their Ru(η6-p-cymene)Cl complexes 2a-2f, and report here the molecular structures of
Oh Kyung Choi et al.
Journal of hazardous materials, 403, 123636-123636 (2020-08-28)
Solvent extraction desalination (SED) is one of the liquid-liquid separation techniques that selectively uptake freshwater from high saline water, and then separate the absorbed freshwater from the solvent through temperature swing. This study evaluated the desalination performance of seven different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service