Skip to Content
Merck
All Photos(4)

Key Documents

125598

Sigma-Aldrich

1,4-Cyclohexanedimethanol

mixture of cis and trans, 99%

Synonym(s):

1,4-Bis(hydroxymethyl)cyclohexane, mixture of cis and trans

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH2OH)2
CAS Number:
Molecular Weight:
144.21
Beilstein:
1902271
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5 (vs air)

Assay

99%

autoignition temp.

584 °F

bp

283 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1CCC(CO)CC1

InChI

1S/C8H16O2/c9-5-7-1-2-8(6-10)4-3-7/h7-10H,1-6H2

InChI key

YIMQCDZDWXUDCA-UHFFFAOYSA-N

Related Categories

General description

1,4-Cyclohexanedimethano is extensively used as cross-linking reagent in polymer industry.

Application

1,4-Cyclohexanedimethanol has been used in the synthesis of polyketal copolymers. It was used as diol comonomer during the synthesis of polyester-carbonates based on 1,3-propylene-co-1,4-cyclohexanedimethylene succinate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New biodegradable polymers from renewable sources: Polyester-carbonates based on 1, 3-propylene-co-1, 4-cyclohexanedimethylene succinate.
Liu Y, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 39(14), 2508-2519 (2001)
Stephen C Yang et al.
Bioconjugate chemistry, 19(6), 1164-1169 (2008-05-27)
Acute inflammatory diseases are a major cause of death in the world, and effective treatments are greatly needed. Macrophages play a central role in causing acute inflammatory diseases, and there is currently great interest in developing drug delivery vehicles that
Mingliang Ding et al.
Biodegradation, 23(1), 127-132 (2011-07-07)
Enzymatic hydrolytic degradation of polybutylene succinate (PBS), poly(polybutylenesuccinate-co-1,4-cyclohexane dimethanol) (PBS/CHDM) and poly(polybutylene succinate-co-diglycolic acid) (PBS/DGA) in mixed solvent of tetrahydrofuran (THF) and toluene was examined. Lipase was used as catalyst to degrade polymers with molecular weight of more than 100,000
Zehong Xiang et al.
Macromolecular bioscience, 19(8), e1900112-e1900112 (2019-06-22)
Inflammation-associated thrombosis is a non-negligible source of mortalities and morbidities worldwide. To manipulate inflammation-associated coagulation, nanoparticles that contain anti-inflammatory polymer (copolyoxalate containing vanillyl alcohol, PVAX) and anti-thrombotic heparin derivative deoxycholic acid (Hep-DOCA) are prepared. The strategy takes advantage of the
Alexandre Trogé et al.
The Journal of the Acoustical Society of America, 128(5), 2704-2714 (2010-11-30)
This paper describes the acoustic properties of a range of epoxy resins prepared by photocuring that are suitable for application in piezoelectric ultrasonic transducer matching layers. Materials, based on blends of diglycidyl ether of Bisphenol A and 1,4-cyclohexanedimethanol diglycidyl ether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service