Skip to Content
Merck
All Photos(1)

Key Documents

124672

Sigma-Aldrich

10-Undecenoic acid

98%

Synonym(s):

Undecylenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8COOH
CAS Number:
Molecular Weight:
184.28
Colour Index Number:
42650
Beilstein:
1762631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

refractive index

n20/D 1.449 (lit.)

bp

137 °C/2 mmHg (lit.)

mp

23-25 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble
chloroform: soluble
diethyl ether: soluble

density

0.912 g/mL at 25 °C (lit.)

functional group

allyl
carboxylic acid

SMILES string

OC(=O)CCCCCCCCC=C

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)

InChI key

FRPZMMHWLSIFAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

10-Undecenoic acid undergoes copolymerisation with ethylene using the metallocene catalyst system Cp2ZrCl2 / methylaluminoxane[1].

Application

10-Undecenoic acid has been used to study the methods and mechanisms for the formation of molecular monolayers on silicon surface[2]. It was used as starting reagent in the syntheses of pheromone (11Z)-hexadecenal and agriculturally important insect pheromones[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metallocene-Catalysed Copolymerisation of Ethylene with 10-Undecenoic Acid: The Effect of Experimental Conditions.
Santos JM, et al.
Macromolecular Chemistry and Physics, 202(11), 2195-2201 (2001)
10-Undecenoic acid, an inexpensive source for the synthesis of the pheromones of cotton pests, peach tree borer and cherry tree borer.
Pawar AS and Chattopadhyay S.
Molecules (Basel), 2(6), 87-90 (1997)
Molecular monolayers on silicon surfaces.
Lopinski GP and Wayner DDM.
Material Matters, 3(2), 38-38 (2008)
Bin Guan et al.
Scientific reports, 5, 12641-12641 (2015-08-01)
This Report presents a nitrogen-doping method by chemically forming self-assembled monolayers on silicon. Van der Pauw technique, secondary-ion mass spectroscopy and low temperature Hall effect measurements are employed to characterize the nitrogen dopants. The experimental data show that the diffusion
Betar M Gallant et al.
ACS nano, 9(5), 5143-5153 (2015-04-16)
The interfacial shear strength between Si microwires and a Nafion membrane has been tailored through surface functionalization of the Si. Acidic (-COOH-terminated) or basic (-NH2-terminated) surface-bound functionality was introduced by hydrosilylation reactions to probe the interactions between the functionalized Si

Articles

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Questions

  1. Is this product in liquid form? If so what is in it that allows it to be liquid at room temperature?

    1 answer
    1. The melting point of this material is 23-25 °C. The physical form will depend on the ambient temperature. Warming to reliquify will not be detrimental to the compound.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service