Skip to Content
Merck
All Photos(1)

Key Documents

123617

Sigma-Aldrich

Benzo[h]quinoline

97%

Synonym(s):

1-Naphthoquinoline, 7,8-Benzoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9N
CAS Number:
Molecular Weight:
179.22
Beilstein:
120249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

338 °C/719 mmHg (lit.)

mp

48-50 °C (lit.)

SMILES string

c1ccc2c(c1)ccc3cccnc23

InChI

1S/C13H9N/c1-2-6-12-10(4-1)7-8-11-5-3-9-14-13(11)12/h1-9H

InChI key

WZJYKHNJTSNBHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzo[h]quinoline was used to study the mutagenic activities of benzo[f]quinoline, benzo[h]quinolone and a number of their derivatives in strain TA 100 of Salmonella typhimurium. It was used in determination of nitrogen-containing polynuclear aromatic hydrocarbons in the gaseous products of the thermal degradation of polymers by HPLC- fluorescence detection. It was used as starting reagent for the synthesis of osmium and ruthenium complexes containing an N-heterocyclic carbene ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J LaVoie et al.
Japanese journal of cancer research : Gann, 78(2), 139-143 (1987-02-01)
The environmental occurrence and mutagenic activity of quinoline and benzoquinolines are well-documented. In this study, the relative carcinogenic activities of quinoline, benzo[f]quinoline, benzo[h]quinoline, and phenanthridine were evaluated in newborn mice. Mice were injected intraperitoneally on the first, eighth, and fifteenth
M Wilhelm et al.
Journal of chromatography. A, 878(2), 171-181 (2000-06-24)
A method for the simultaneous determination of 22 nitrogen-containing polynuclear aromatic hydrocarbons (PAHs) (15 azaarenes and seven amino-PAHs) in the gaseous products of the thermal degradation of polymers is described. After desorption and clean-up using cation-exchange chromatography (PRS cartridge) the
E C Riesgo et al.
Inorganic chemistry, 40(14), 3413-3422 (2001-06-26)
The Friedländer condensation was employed to synthesize two series of 3,3'-polymethylene bridged ligands, L, based on 2-(2'-pyridyl)-benzo[h]quinoline and 2,2'-bibenzo[h]quinoline (BHQ) along with the fully aromatic naphtho[1,2-b]-1,10-phenanthroline. Complexes [Cu(L)(2)](+) were prepared as their perchlorate or hexafluorophosphate salts. The solution state structures
Hanumantharao Paritala et al.
Bioorganic & medicinal chemistry letters, 19(6), 1584-1587 (2009-02-27)
G-quadruplexes are unusual structures formed from guanine-rich sequences of nucleic acids. G-quadruplexes have been postulated to play important roles in a number of biological systems including gene regulation and the inhibition of enzyme function. Recently, our laboratory reported on the
John B Sutherland et al.
Applied microbiology and biotechnology, 67(3), 405-411 (2005-04-28)
Cultures of Umbelopsis ramanniana (=Mucor ramannianus) were grown in fluid Sabouraud medium for 3 days, dosed with 0.23 mM benzo[f]quinoline, benzo[h]quinoline, or phenanthridine (benzo[c]quinoline), and incubated for another 18 days. Cultures were extracted and metabolites (66-75% of the UV absorbance)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service