123250
4-Antipyrinecarboxaldehyde
97%
Synonym(s):
2,3-Dimethyl-5-oxo-1-phenyl-3-pyrazoline-4-carboxaldehyde, Antipyraldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H12N2O2
CAS Number:
Molecular Weight:
216.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
162-165 °C (lit.)
functional group
aldehyde
SMILES string
[H]C(=O)C1=C(C)N(C)N(c2ccccc2)C1=O
InChI
1S/C12H12N2O2/c1-9-11(8-15)12(16)14(13(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3
InChI key
QFYZFYDOEJZMDX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Antipyrinecarboxaldehyde undergoes condensation with 4′-aminobenzo-15-crown-5- to yield functionalized crown ether.
Application
4-Antipyrinecarboxaldehyde was used in the synthesis of a new crown ether-antipyrine schiffs base.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ashraf S Hassan et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-06)
A series of Bis-pyrazole Schiff bases (6a-d and 7a-d) and mono-pyrazole Schiff bases (8a-d and 9a-d) were designed and synthesized through the reaction of 5-aminopyrazoles 1a-d with aldehydes 2-5 using mild reaction condition with a good yield percentage. The chemical
Preparation and spectroscopic characterization of iron (II) and copper (II) complexes of a functionalized crown ether.
Hayvali Z.
Transition Metal Chemistry, 34(1), 97-101 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service