Skip to Content
Merck
All Photos(2)

Documents

122629

Sigma-Aldrich

Ethyl levulinate

greener alternative

99%

Synonym(s):

Ethyl 4-oxopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COOC2H5
CAS Number:
Molecular Weight:
144.17
Beilstein:
507641
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.422 (lit.)

bp

93-94 °C/18 mmHg (lit.)

solubility

H2O: freely soluble
alcohol: miscible

density

1.016 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CCOC(=O)CCC(C)=O

InChI

1S/C7H12O3/c1-3-10-7(9)5-4-6(2)8/h3-5H2,1-2H3

InChI key

GMEONFUTDYJSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl levulinate is diluent for biodiesel fuels with high saturated fatty acid content.

Ethyl levulinate is a ketoester produced through the esterification of levulinic acid and is used as an oxygenated diesel additive.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been classified as a greener solvent. Click here for more information.

Application

Ethyl levulinate was used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ethyl levulinate: A potential bio-based diluent for biodiesel which improves cold flow properties.
Joshi H, et al
Biomass and Bioenergy, 35(7), 3262-3266 (2011)
Optimized enzymatic synthesis of levulinate ester in solvent-free system
Alice L et al.
Industrial Crops and Products, 32, 246-251 (2010)
T Hirata
International journal of leprosy and other mycobacterial diseases : official organ of the International Leprosy Association, 47(4), 585-596 (1979-12-01)
The fine structures and the interconnections between the intracytoplasmic membrane systems and cell division of murine leprosy bacilli in liver tissue from mice infected intraperitoneally with Mycobacterium lepraemurium, Hawaiian strain, were studied in ultrathin serial sections at the electron microscopic
Catalytic upgrading of levulinic acid to ethyl levulinate using reusable silica-included Wells-Dawson heteropolyacid as catalyst.
Pasquale G, et al.
Catalysis Communications, 18, 115-120 (2012)
Bénédicte Pineau et al.
Journal of agricultural and food chemistry, 57(9), 3702-3708 (2009-03-31)
A preparative HPLC method, which preserves wine aromas and isolates fruity characteristics in specific fractions, was applied to red wine aroma extracts. Various odor-active zones were detected in typical fractions by GC-O analysis of their extracts. Through further GC-MS analyses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service