Skip to Content
Merck
All Photos(1)

Key Documents

118184

Sigma-Aldrich

5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans

≥99%

Synonym(s):

Isophorone diamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H7(CH3)3CH2NH2
CAS Number:
Molecular Weight:
170.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

refractive index

n20/D 1.488 (lit.)

bp

247 °C (lit.)

mp

10 °C (lit.)

density

0.922 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CC(N)CC(C)(CN)C1

InChI

1S/C10H22N2/c1-9(2)4-8(12)5-10(3,6-9)7-11/h8H,4-7,11-12H2,1-3H3

InChI key

RNLHGQLZWXBQNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans (IPD), is an cyclo-aliphatic diamine that is mainly used as an epoxy curing agent.

Application

IPD is widely used as a hardener to cure a variety of epoxy based resins, such as diglycidyl ether bisphenol A (DGEBA), and diglycidyl ether 1,4-butanediol (DGEBU)..

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Durability of adhesives based on different epoxy/aliphatic amine networks
Garcia F, et al.
International Journal of Adhesion and Adhesives, 31(4), 177-181 (2011)
Divanillin-Based Epoxy Precursors as DGEBA Substitutes for Biobased Epoxy Thermosets
Savonnet E, et al.
ACS sustainable chemistry & engineering, 6(8), 11008-11017 (2018)
Effect of amine-functionalized silica nanoparticles on thermal and mechanical behaviors of DGEBA/IPD epoxy networks
Behniafar H and Nazemi MK
Polymer Bull., 74(9), 3739-3749 (2017)
High enthalpy storage thermoset network with giant stress and energy output in rubbery state
Fan J and Li G
Nature Communications, 9(1), 642-642 (2018)
Klaus Becker et al.
PloS one, 9(12), e114149-e114149 (2014-12-03)
Tissue clearing allows microscopy of large specimens as whole mouse brains or embryos. However, lipophilic tissue clearing agents as dibenzyl ether limit storage time of GFP-expressing samples to several days and do not prevent them from photobleaching during microscopy. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service