Skip to Content
Merck
All Photos(1)

Key Documents

111309

Sigma-Aldrich

1-Phenyl-1-propanol

≥97%

Synonym(s):

α-Ethylbenzyl alcohol, (±)-1-Phenylpropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(C6H5)OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
1906759
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023126
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

refractive index

n20/D 1.52 (lit.)

bp

103 °C/14 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

CCC(O)c1ccccc1

InChI

1S/C9H12O/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3

InChI key

DYUQAZSOFZSPHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Phenyl-1-propanol enantiomers (E-PP) forms the inclusion complexes of cyclic decapeptide (CDP) which were studied using the density functional theory (DFT) B3LYP method. It was also used to determine the mass transfer kinetics effect on the elution profiles of the 1-phenyl-1-propanol (PP) enantiomers on Chiracel OB (cellulose tribenzoate coated on silica gel).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Arvind Rajendran et al.
Journal of chromatography. A, 1092(1), 55-64 (2005-09-29)
The enantioseparation of 1-phenyl-1-propanol through the supercritical fluid-simulated moving bed (SF-SMB) process is studied. Non-linear isotherms were measured on an analytical column, and used together with the triangle theory for SMB design to select operating conditions for the SF-SMB. Experiments
Pieter Saveyn et al.
Langmuir : the ACS journal of surfaces and colloids, 25(19), 11322-11327 (2009-09-03)
The influence of the physicochemical properties of the vesicular bilayer on the sorption of poorly water soluble compounds was investigated with pulsed field gradient 1H nuclear magnetic resonance (PFG-NMR) for the case of phosphatidylcholine and dioctadecyldimethylammonium bromide (DODAB), using 4-ethylbenzyl
Stefan Ottiger et al.
Journal of chromatography. A, 1162(1), 74-82 (2007-02-17)
The separation of the enantiomers of 1-phenyl-1-propanol by supercritical fluid chromatography on a chiral stationary phase, which consists of cellulose tris (3,5-dimethylphenylcarbamate) coated on a silica support (Chiralcel-OD), is studied under overloaded, non-linear chromatographic conditions. Pulse experiments are performed at
Hongge Zhao et al.
Journal of molecular modeling, 18(3), 851-858 (2011-06-01)
Cyclic peptides are exciting novel hosts for chiral and molecular recognition. In this work, the inclusion complexes of cyclic decapeptide (CDP) with the 1-phenyl-1-propanol enantiomers (E-PP) are firstly studied using the density functional theory (DFT) B3LYP method. Our calculated results
S Khattabi et al.
Journal of chromatography. A, 877(1-2), 95-107 (2000-06-14)
Using single-step frontal analysis, we measured single-component and competitive adsorption isotherm data for the two enantiomers of 1-phenyl-1-propanol (PP). These experimental data were fitted to several competitive bi-Langmuir models (with 8, 6, 5 and 4 parameters) and to the competitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service