Skip to Content
Merck
All Photos(1)

Key Documents

110752

Sigma-Aldrich

Ethylcyclopentane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9C2H5
CAS Number:
Molecular Weight:
98.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

72.8 mmHg ( 37.7 °C)

Assay

98%

autoignition temp.

500 °F

expl. lim.

6.7 %

refractive index

n20/D 1.419 (lit.)

bp

103 °C (lit.)

mp

−138 °C (lit.)

density

0.763 g/mL at 25 °C (lit.)

SMILES string

CCC1CCCC1

InChI

1S/C7H14/c1-2-7-5-3-4-6-7/h7H,2-6H2,1H3

InChI key

IFTRQJLVEBNKJK-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis A Rios-Hernandez et al.
Applied and environmental microbiology, 69(1), 434-443 (2003-01-07)
We used ethylcyclopentane (ECP) as a model alicyclic hydrocarbon and investigated its metabolism by a sulfate-reducing bacterial enrichment obtained from a gas condensate-contaminated aquifer. The enrichment coupled the consumption of ECP with the stoichiometrically expected amount of sulfate reduced. During
Gary B McVicker et al.
The journal of physical chemistry. B, 109(6), 2222-2226 (2006-07-21)
In this paper we describe the utility of an acid-catalyzed isomerization reaction, specifically, ring-contraction of methylcyclohexane to an isomeric mixture of alkylcyclopentanes as a tool for characterizing the acidic properties of a wide range of platinum-loaded solid acids. Methylcyclohexane isomerization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service