Skip to Content
Merck
All Photos(1)

Key Documents

110205

Sigma-Aldrich

(±)-Propylene oxide

ReagentPlus®, ≥99%

Synonym(s):

(±)-Methyloxirane, 1,2-Epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein:
79763
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

2 (vs air)

Quality Level

vapor pressure

29.43 psi ( 55 °C)
8.59 psi ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

autoignition temp.

1378 °F

expl. lim.

37 %

refractive index

n20/D 1.366 (lit.)

bp

34 °C (lit.)

mp

−112 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3

InChI key

GOOHAUXETOMSMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-Propylene oxide is an organic compound used as an intermediate in several chemical reactions. It readily reacts with active hydrogen atoms.

Application

Used in the production of polyether polyols, propylene glycols, propylene glycol ethers.Used as an intermediate to produce ammonium type complex to remove thin films of petroleum from water surfaces.

Other Notes

The article number 110205-4X1L will be discontinued. Please order the single bottle 110205-1L which is physically identical with the same exact specifications.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-36.4 °F - Equilibrium method

Flash Point(C)

-38 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josep Casamada Ribot et al.
Chemical communications (Cambridge, England), 46(37), 6971-6973 (2010-08-24)
The aqueous gelation of a quaternary ammonium oligo(propylene oxide)-based ionic liquid yields ion gels with a reverse thermoresponsive behavior (i.e., mechanical moduli and viscosity increase with temperature) and enhanced ionic conductivities.
Javix Thomas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(16), 4582-4587 (2011-03-25)
Chirality recognition in the hydrogen-bonded glycidol···propylene oxide complex has been studied by using rotational spectroscopy and ab initio calculations. An extensive conformational search has been performed for this binary adduct at the MP2/6-311++G(d,p) level of theory and a total of
Jasper Jammaer et al.
Journal of the American Chemical Society, 133(34), 13737-13745 (2011-07-28)
Hexagonally ordered mesoporous silica coined COK-12 was synthesized in a continuous process by combining streams of sodium silicate and citric acid/sodium citrate buffered solution of (ethylene oxide)(20)-(propylene oxide)(70)-(ethylene oxide)(20) triblock copolymer (Pluronic P123) from separate reservoirs. COK-12 precipitated spontaneously upon
Christian M Hammer et al.
Journal of cataract and refractive surgery, 41(6), 1279-1288 (2015-07-21)
To assess the suitability of a new 345 nm ultraviolet (UV) femtosecond laser for refractive surgery. Department of Ophthalmology, University of Erlangen-Nürnberg, Erlangen, Germany. Experimental study. Twenty-five porcine corneas were used for stromal flap or lamellar bed creation (stromal depth
Satoshi Toda et al.
Blood, 123(25), 3963-3971 (2014-03-25)
Definitive erythropoiesis takes place at erythroblastic islands, where erythroblasts proliferate and differentiate in association with central macrophages. At the final stage of erythropoiesis, pyrenocytes (nuclei surrounded by plasma membranes) are excluded from erythroblasts, expose phosphatidylserine (PtdSer), and are engulfed by

Questions

  1. Is it suitable for electron microscopy processing?

    1 answer
    1. Suitability of this product with electron microscopy is not tested. It may be suitable, but the customer would need to ensure suitability. A propylene oxide which is tested with electron microscopy is not offered.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service