Skip to Content
Merck
All Photos(1)

Key Documents

108111

Sigma-Aldrich

Benzenesulfonyl chloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2Cl
CAS Number:
Molecular Weight:
176.62
Beilstein:
606926
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.04 mmHg ( 20 °C)

Quality Level

Assay

96%

refractive index

n20/D 1.551 (lit.)

bp

251-252 °C (lit.)

mp

13-15 °C (lit.)

solubility

alcohol: soluble
cold water: insoluble
diethyl ether: soluble

density

1.384 g/mL at 25 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccccc1

InChI

1S/C6H5ClO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H

InChI key

CSKNSYBAZOQPLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzenesulfonyl chloride reacts with Grignard reagent from N-unsubstituted indoles to form oxindoles or substituted indoles. It is the derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry.

Application

Benzenesulfonyl chloride may be used in thiamine assay for determination of thiamine in different food products.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

260.6 °F

Flash Point(C)

127 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unusual reactions of magnesium indolates with benzenesulfonyl chloride.
Wenkert E, et al.
The Journal of Organic Chemistry, 52(15), 3404-3409 (1987)
A Leone-Bay et al.
Journal of medicinal chemistry, 38(21), 4257-4262 (1995-10-13)
A series of benzoylated and phenylsulfonylated amino acids are novel, low molecular weight, self-assembling molecules. At low pH, these compounds form microspheres that dissolve readily under neutral conditions. In a given synthetic series, those molecules with low aqueous solubility formed
Ihor E Kopka et al.
Bioorganic & medicinal chemistry letters, 12(4), 637-640 (2002-02-15)
A series of substituted N-(3,5-dichlorobenzenesulfonyl)-L-prolyl- and alpha-methyl-L-prolyl-phenylalanine derivatives was prepared as VLA-4/VCAM antagonists. The compounds showed excellent potency with a wide variety of neutral, polar, electron withdrawing or donating groups on the phenylalanine ring (IC50 approximately 1 nM). Heteroaryl ring
A G Soliman
Journal - Association of Official Analytical Chemists, 64(3), 616-622 (1981-05-01)
A semiautomated procedure was used to measure the fluorescence of sample extracts before and after the addition of benzenesulfonyl chloride (BSC). Addition of BSC inhibited thiochrome formation and provided a more representative blank based on the fluorescence of all the
Analysis of primary and secondary aliphatic amines in waste water and surface water by gas chromatography-mass spectrometry after derivatization with 2, 4-dinitrofluorobenzene or benzenesulfonyl chloride.
Sacher F, et al.
Journal of Chromatography A, 764(1), 85-93 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service