Skip to Content
Merck
All Photos(1)

Documents

10445

Sigma-Aldrich

Anisaldehyde dimethyl acetal

≥98.5% (GC)

Synonym(s):

4-Methoxybenzaldehyde dimethyl acetal, α,α,4-Trimethoxytoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein:
2048930
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5% (GC)

refractive index

n20/D 1.505

bp

85-87 °C/0.1 mmHg (lit.)

solubility

H2O: soluble
alcohol: soluble

density

1.07 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(OC)c1ccc(OC)cc1

InChI

1S/C10H14O3/c1-11-9-6-4-8(5-7-9)10(12-2)13-3/h4-7,10H,1-3H3

InChI key

NNHYAHOTXLASEA-UHFFFAOYSA-N

General description

Anisaldehyde dimethyl acetal is a clear, colorless to light yellow liquid. The flash point for anisaldehyde dimethyl acetal is 114.0°C. It undergoes allylation reactions with allyltrimethylsilane catalysed by Iron(III) chloride. Anisaldehyde dimethyl acetal undergoes ytterbium-catalyzed tandem carboalkoxylation/Friedel–Crafts reaction with 1-phenylbenzylidenecyclopropane to yield 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene.

Application

Anisaldehyde dimethyl acetal was used in the synthesis of lipid A mimic precursor. It was also used in the synthesis of 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate.

Other Notes

Protecting group reagent for diols, used e.g. in carbohydrate chemistry; the acetal protection can be selectively cleaved under mild conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Kloosterman et al.
Rec. Trav. Chim., 103, 243-243 (1984)
R. Johansson et al.
Journal of the Chemical Society. Perkin Transactions 1, 2371-2371 (1984)
Synthesis of a Precursor of a Lipid A Mimic.
Barath M, et al.
Chemical Papers, 57(2), 125-130 (2003)
Iron (III) chloride-catalyzed effective allylation reactions of aldehydes with allyltrimethylsilane.
Watahiki T and Oriyama T.
Tetrahedron Letters, 43(49), 8959-8962 (2002)
Yasumasa Hara et al.
Journal of natural medicines, 72(1), 280-289 (2017-12-07)
A new amide, named dehydropropylpantothenamide (1), was obtained by a co-culture of Nocardia tenerifensis IFM 10554

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service