Skip to Content
Merck
All Photos(1)

Key Documents

103764

Sigma-Aldrich

Pentafluorobenzenesulfonyl chloride

99%

Synonym(s):

2,3,4,5,6-Pentafluorophenylsulfonyl chloride, Perfluorobenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5SO2Cl
CAS Number:
Molecular Weight:
266.57
Beilstein:
2142325
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

refractive index

n20/D 1.479 (lit.)

bp

210-211 °C (lit.)

density

1.796 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Fc1c(F)c(F)c(c(F)c1F)S(Cl)(=O)=O

InChI

1S/C6ClF5O2S/c7-15(13,14)6-4(11)2(9)1(8)3(10)5(6)12

InChI key

UOJCTEGNHXRPKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentafluorobenzenesulfonyl chloride reacts with benzene and thiophene derivatives in the presence of a ruthenium (II) catalyst to form corresponding perfluorophenylated compounds.

Application

Pentafluorobenzenesulfonyl chloride was used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. Pentafluorobenzenesulfonyl chloride was used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection.

Biochem/physiol Actions

Pentafluorobenzenesulfonyl chloride is derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detectio.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perfluorophenylation of aromatic and heteroaromatic compounds with pentafluorobenzenesulfonyl chloride catalyzed by a ruthenium (II) phosphine complex.
Kamigata N, et al.
Journal of Fluorine Chemistry, 87(1), 91-95 (1998)
Pentafluorobenzenesulfonyl chloride as a sensitive reagent for the rapid gas chromatographic analysis of tranylcypromine in tissues and body fluids.
T S Rao et al.
Biochemical pharmacology, 35(12), 1925-1928 (1986-06-15)
S J Asghar et al.
Journal of pharmacological and toxicological methods, 46(2), 111-115 (2002-12-17)
Acute administration of (+)-amphetamine has been used as a model for mania in humans since it mimics the physiological, biochemical, and cognitive effects seen in mania. A rapid and sensitive method for the determination of amphetamine in human plasma samples
Pentafluorobenzenesulfonyl chloride: a new electrophoric derivatizing reagent with application to tyrosyl peptide determination by gas chromatography with electron capture detection.
A Sentissi et al.
Analytical chemistry, 56(13), 2512-2517 (1984-11-01)
G B Baker et al.
Journal of chromatography, 381(2), 211-217 (1986-09-05)
A gas chromatographic procedure is described for the analysis of beta-phenylethylamine (PEA) in tissues and body fluids. The method involves the use of pentafluorobenzenesulfonyl chloride for extraction and derivatization of PEA. This is followed by separation and analysis of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service