Skip to Content
Merck
All Photos(2)

Key Documents

103713

Sigma-Aldrich

2,3,4,5,6-Pentafluoroaniline

99%

Synonym(s):

Pentafluoroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5NH2
CAS Number:
Molecular Weight:
183.08
Beilstein:
1819387
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

153 °C (lit.)

mp

33-35 °C (lit.)

solubility

toluene: soluble

functional group

fluoro

SMILES string

Nc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C6H2F5N/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H2

InChI key

NOXLGCOSAFGMDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,4,5,6-Pentafluoroaniline forms metal-drug complexes, cis-Pt-(2,3,4,5,6-pentafluoroaniline)2-Br2 which has been tested against the promastigote forms of Leishmania donovani.

Application

2,3,4,5,6-Pentafluoroaniline may be used in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. 2,3,4,5,6-Pentafluoroaniline was used in synthesis of various titanium complexes having two anionic [N, O] bidentate salicylaldiminato ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

163.4 °F

Flash Point(C)

73 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentafluorophenylammonium triflate (PFPAT): an efficient, practical, and cost-effective catalyst for esterification, thioesterification, transesterification, and macrolactone formation.
Funatomi T, et al.
Green Chemistry, 8(12), 1022-1027 (2006)
Bis (salicylaldiminato) titanium complexes containing bulky imine substituents: Synthesis, characterization and ethene polymerization studies.
Parssinen A, et al.
European Journal of Inorganic Chemistry, 2005(11), 2100-2109 (2005)
C M Mesa-Valle et al.
The Journal of antimicrobial chemotherapy, 40(1), 47-57 (1997-07-01)
The action of 16 newly synthesized metal complexes having the general structure cis-Pt-(II)-Xn-Ln have been tested in vitro against the promastigote forms of Leishmania donovani. The metal complexes at 24 h and maximum dosages inhibited growth from 0%, e.g. in
Vasily A Ilichev et al.
Dalton transactions (Cambridge, England : 2003), 48(3), 1060-1066 (2019-01-03)
To obtain new efficient lanthanide-based NIR luminophores perfluorinated 2-mercaptobenzothiazole was used as a ligand. The ate-complexes [(Ln(mbtF)4)-(Na(DME)3)+] of Nd (1), Sm (2), Tb (3), Er (4), Yb (5) and [(Y(mbtF)4)-(Li(DME)3)+] (6) were synthesized in high yields by the reactions of
I M Rietjens et al.
Chemico-biological interactions, 77(3), 263-281 (1991-01-01)
Metabolism and bioactivation of fluoroanilines was studied both in vitro in microsomal systems and in vivo. 4-Fluoroaniline and pentafluoroaniline and their non-para fluorinated analogues were used as the model compounds. Special attention was focussed on bioactivation to reactive benzoquinoneimines. Cytochrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service