Skip to Content
Merck
All Photos(1)

Key Documents

101656

Sigma-Aldrich

2-Chloro-4-nitroaniline

99%

Synonym(s):

1-Amino-2-chloro-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)NH2
CAS Number:
Molecular Weight:
172.57
Beilstein:
638657
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

autoignition temp.

971 °F

mp

105-108 °C (lit.)

SMILES string

Nc1ccc(cc1Cl)[N+]([O-])=O

InChI

1S/C6H5ClN2O2/c7-5-3-4(9(10)11)1-2-6(5)8/h1-3H,8H2

InChI key

LOCWBQIWHWIRGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Borcherding et al.
Archives of environmental contamination and toxicology, 40(4), 497-504 (2001-08-30)
The Dreissena-Monitor is a biological early warning system for the continuous monitoring of river water quality, based on the valve movements of two groups of 42 zebra mussels (Dreissena polymorpha). Laboratory experiments with Cd, PCP, and 2-chloro-4-nitro-aniline were conducted in
Thai Ha Tran et al.
Chemosphere, 201, 425-431 (2018-03-13)
The manganese oxide birnessite adsorbed and catalyzed the transformation of the anthelminthic drug niclosamide (NIS) into 2-chloro-4-nitroaniline (CNA) and 5-chlorosalicylic acid (CSA) at acidic pH. The adsorption of NIS was fitted using a linear isotherm for all conditions and reaction
Jimmy Murillo-Gelvez et al.
Environmental science & technology, 53(10), 5816-5827 (2019-05-01)
Nitroaromatic compounds (NACs) are a class of prevalent contaminants. Abiotic reduction is an important fate process that initiates NAC degradation in the environment. Many linear free energy relationship (LFER) models have been developed to predict NAC reduction rates. Almost all
J J Espinosa-Aguirre et al.
Mutation research, 264(3), 139-145 (1991-11-01)
Niclosamide is an anti-helminthic drug susceptible to being metabolized into a bacterial mutagen by the action of enzymes present in the S9 activation mixture. Additional results from genotoxic studies in rodents and humans suggest that the drug is absorbed from
A L Scher et al.
Journal of AOAC International, 76(2), 287-291 (1993-03-01)
A method is described for the determination of the intermediates and a related impurity in D&C Red No. 36 by reversed-phase liquid chromatography. This method may be used to ensure that limits set forth in the Code of Federal Regulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service