Merck
  • Determination of high-affinity choline uptake (HACU) and choline acetyltransferase (ChAT) activity in the same population of cultured cells.
Determination of high-affinity choline uptake (HACU) and choline acetyltransferase (ChAT) activity in the same population of cultured cells.
Brain research (2009-08-08)
Balmiki Ray, Jay R Simon, Debomoy K Lahiri
ABSTRACT
Cholinergic neurons are a major constituent of the mammalian central nervous system. Acetylcholine, the neurotransmitter used by cholinergic neurons, is synthesized from choline and acetyl CoA by the enzymatic action of choline acetyltransferase (ChAT). The transport of choline into the cholinergic neurons, which results in synthesis of ACh, is hemicholinium-sensitive and is referred to as high-affinity choline uptake (HACU). Thus, the formation of acetylcholine in cholinergic neurons largely depends on both the levels of choline being transported into the cells from the extracellular space and the activity of ChAT. Several methods were described previously to measure HACU and ChAT simultaneously in synaptosomes, but the same for cultured cells is lacking. We describe a procedure to measure HACU and ChAT at the same time in cultured cells by simple techniques employing radionuclides. In this procedure, we determined quantitatively hemicholinium-sensitive choline uptake and ChAT enzyme activity in a small number of differentiated human neuroblastoma (SK-N-SH) cells. We also determined the kinetics of choline uptake in the SK-N-SH cells. We believe that these simple methods can be used for neurochemical and drug discovery studies in several models of neurodegenerative disorders including Alzheimer's disease.
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Hemicholinium-3, solid, ≥95% (HPLC)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, Molecular Biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Supelco
trans-3′-Hydroxycotinine-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%