Merck
  • Loss of function CHCHD10 mutations in cytoplasmic TDP-43 accumulation and synaptic integrity.
Loss of function CHCHD10 mutations in cytoplasmic TDP-43 accumulation and synaptic integrity.
Nature communications (2017-06-07)
Jung-A A Woo, Tian Liu, Courtney Trotter, Cenxiao C Fang, Emillio De Narvaez, Patrick LePochat, Drew Maslar, Anusha Bukhari, Xingyu Zhao, Andrew Deonarine, Sandy D Westerheide, David E Kang
ABSTRACT
Although multiple CHCHD10 mutations are associated with the spectrum of familial and sporadic frontotemporal dementia-amyotrophic lateral sclerosis (FTD-ALS) diseases, neither the normal function of endogenous CHCHD10 nor its role in the pathological milieu (that is, TDP-43 pathology) of FTD/ALS have been investigated. In this study, we made a series of observations utilizing Caenorhabditis elegans models, mammalian cell lines, primary neurons and mouse brains, demonstrating that CHCHD10 normally exerts a protective role in mitochondrial and synaptic integrity as well as in the retention of nuclear TDP-43, whereas FTD/ALS-associated mutations (R15L and S59L) exhibit loss of function phenotypes in C. elegans genetic complementation assays and dominant negative activities in mammalian systems, resulting in mitochondrial/synaptic damage and cytoplasmic TDP-43 accumulation. As such, our results provide a pathological link between CHCHD10-associated mitochondrial/synaptic dysfunction and cytoplasmic TDP-43 inclusions.
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Tdp43 Antibody, clone 3H8, clone 3H8, from mouse