Merck
  • Three-dimensional testicular organoid: a novel tool for the study of human spermatogenesis and gonadotoxicity in vitro.
Three-dimensional testicular organoid: a novel tool for the study of human spermatogenesis and gonadotoxicity in vitro.
Biology of reproduction (2017-03-25)
Samuel S Pendergraft, Hooman Sadri-Ardekani, Anthony Atala, Colin E Bishop
ABSTRACT
Existing methods for evaluating the potential gonadotoxicity of environmental agents and pharmaceutical compounds rely heavily on animal studies. The current gold standard in vivo functional assays in animals are limited in their human predictive capacity. In addition, existing human two-dimensional in vitro models of testicular toxicity do not accurately reflect the in vivo situation. A more reliable testicular in vitro model system is needed to better assess the gonadotoxic potential of drugs prior to progression into clinical trials. The overall goal of this study was to develop a three-dimensional (3D) in vitro human testis organoid culture system for use as both a predictive first tier drug-screening tool and as a model of human testicular function. Multicellular human testicular organoids composed of Spermatogonial Stem Cells, Sertoli, Leydig and peritubular cells were created and evaluated over time for morphology, viability, androgen production and ability to support germ cell differentiation. Enzyme-linked immunosorbent assay measurements confirmed that the organoids produced testosterone continuously with and without hCG stimulation. Upregulation of postmeiotic genes including PRM1 and Acrosin, detected by quantitative-PCR, digital PCR and Immunofluorescence, indicated the transition of a small percentage of diploid to haploid germ cells. As a novel screening tool for reproductive toxicity, 3D organoids were exposed to four chemotherapeutic drugs, and they responded in a dose-dependent manner and maintained IC50 values significantly higher than 2D cultures. This 3D human testis organoid system has the potential to be used as a novel testicular toxicity-screening tool and in vitro model for human spermatogenesis.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-PRM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ACR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Fluoroshield with DAPI, histology mounting medium