Skip to Content
Merck
All Photos(1)

Key Documents

F6057

Sigma-Aldrich

Fluoroshield with DAPI

liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Fluoroshield with DAPI, liquid

description

histology mounting medium

Quality Level

form

liquid

composition

Sodium Azide, ≤1% (also contains other ingredients)
Tris-HCl

technique(s)

immunofluorescence: suitable

pH range

7.9-8.3

solubility

water: soluble

fluorescence

λex 360 nm; λem 460 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

General description

Fluoroshield with DAPI is an aqueous mounting medium fortified with 4′,6-diamidino-2-phenylindole (DAPI), a nuclear counterstain used as a blue-fluorescent DNA probe. It is an anti-fading agent employed for preserving the fluorescence of tissue and cell smears by preventing rapid photobleaching and minimizing the blinking-mediated phenomena.(1) For use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360 nm and emits at 460 nm, producing a blue fluorescence. RNA is also stained with DAPI.

Application

Fluoroshield with DAPI has been used:
  • as a counterstain in immunocytochemistry to label nuclei of neural progenitor cells[1]
  • as a histology mounting medium for cardiac tissues sections for immunofluorescence studies[2]
  • as mounting solution for immunostained heart sections for nuclei detection[3]

Legal Information

FluoroShield is a trademark of ImmunoBioScience Corp.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extracellular matrix-derived extracellular vesicles promote cardiomyocyte growth and electrical activity in engineered cardiac atria
An M, et al.
Biomaterials, 146(1), 49-59 (2017)
Edgar T Hoorntje et al.
PloS one, 13(8), e0203078-e0203078 (2018-08-31)
Likely pathogenic/pathogenic variants in genes encoding desmosomal proteins play an important role in the pathophysiology of arrhythmogenic right ventricular cardiomyopathy (ARVC). However, for a substantial proportion of ARVC patients, the genetic substrate remains unknown. We hypothesized that plectin, a cytolinker
miR-210 suppresses BNIP3 to protect against the apoptosis of neural progenitor cells
Wang F, et al.
Stem Cell Research, 11(1), 657-667 (2013)
S Hamada et al.
Histochemistry, 79(2), 219-226 (1983-01-01)
DNA-DAPI complexes emit strong bluish white fluorescence when excited by ultraviolet light so that even very small amounts of DNA such as those in mitochondria, chloroplasts, and virus particles can be visualized. Moreover, the staining procedure with DAPI is very
Human umbilical cord tissue-derived mesenchymal stromal cells attenuate remodeling after myocardial infarction by proangiogenic, antiapoptotic, and endogenous cell-activation mechanisms
Nascimento DS, et al.
Stem Cell Research & Therapy, 5(1), 5-5 (2014)

Articles

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

See All

Questions

1–10 of 15 Questions  
  1. How is Fluoroshield (F6057) used with DAPI?

    1 answer
    1. Fluoroshield with DAPI is an aqueous mounting medium designed to preserve the fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of various fluorescent dyes. Additionally, it retains fluorescence during prolonged storage at 4°C in the dark. Importantly, the medium does not contain phenylenediamine, which can destroy immunofluorescence of certain dyes. The mounting medium is fortified with DAPI, a counter-stain for DNA, intended for use in situ hybridization techniques or other methods requiring fluorescence of DNA staining. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI. The intended use of this product includes immunofluorescence and confocal microscopy.

      Reagent:
      Fluoroshield with DAPI is a ready-to-use mounting medium.

      Storage:
      It should be stored at 2-8°C, protected from light, and should not be frozen.

      Procedure:
      - Bring the vial to room temperature.
      - Rinse the slide to be mounted with distilled or deionized water, remove excess water by touching the edges of the slide on a paper towel, and place the slides on a flat surface away from light.
      - Apply 3-4 drops of mounting medium directly on top of the specimen and spread it out evenly, ensuring the tissue is not touched. Excess medium can be removed by touching the edges of the slide on a paper towel.
      - Let it stand at room temperature for about 5 minutes.
      - Apply a cover slip, carefully avoiding air bubbles. If a coverslip is not used, air bubbles will appear in a few days.
      - For long-term storage, it is recommended to store the slide in the dark at 2-8°C.

      Removal of Coverslip:
      The coverslip can be removed before sealing the edges. Soak the slide in warm (37°C) water for a few minutes, carefully and slowly moving.

      Helpful?

  2. What could be the reason for the occurrence of a white film after curing, particularly when the coverslips are slightly wet with PBS?

    1 answer
    1. The following recommendations are provided:

      •If there is tissue attached to glass slides, it is recommended to use a Kimwipe to carefully dry the slide around the tissue section.
      •If cells are on a coverslip, removing excess water from the coverslip by placing it on its end and then using a paper towel to wick away excess moisture is suggested.
      •The possibility that as the slides dry, salts from the PBS are evaporating and leaving a white residue or film is considered.

      Helpful?

  3. What are the potential implications of storing a bottle of F6057-20 at -20°C for 2 days by mistake?

    1 answer
    1. It is suggested to allow the bottle to thaw. Once thawed, the bottle should be inverted several times to thoroughly mix its contents. If the solution looks normal and does not contain any precipitate or particulate matter, it is expected that the mounting media is suitable for use.

      Helpful?

  4. What are the procedures for solubilizing and using Fluoroshield with DAPI (F6057)?

    1 answer
    1. Fluoroshield with DAPI is a solution that is ready to use and does not require solubilization. It is an aqueous mounting medium designed to preserve the fluorescence of tissue and cell smears. The mounting medium is fortified with DAPI, a counter-stain for DNA, and is intended for use in situ hybridization techniques, immunofluorescence and confocal microscopy.

      Helpful?

  5. Is sealing a 96-well plate necessary in the final step of ICC with Fluoroshield™ with DAPI, as done with coverslips using clear nail polish?

    1 answer
    1. Fluoroshield with DAPI is designed for use with glass slides or coverslips intended for analysis using a fluorescent microscope. It is not commonly used for coverslipping 96 well plates. There are no known individual, small, round coverslips for each of the 96 wells, nor are there large clear glass plates to fit over the top of a 96 well plate. If such glass plates are available, it is not known that such a product exists.

      Helpful?

  6. what is the refractive index?

    1 answer
    1. The refractive index is not determined internally for this product. However, the supplier reports that it is 1.364 ± 0.002 of the product in solution.

      Helpful?

  7. Does Fluoroshield (Product Nos. F6182, F5932, F6057) contain an antifade reagent?

    1 answer
    1. This unique formula prevents rapid photobleaching.  The exact nature of the antifade is considered proprietary.

      Helpful?

  8. Why is DAPI (4´,6-Diamidino-2-phenylindole dihydrochloride ) added to Fluoroshield with DAPI mounting media, Product No. F6057?

    1 answer
    1. This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360 nm and emits at 460 nm, producing a blue fluorescence. RNA is also stained with DAPI.

      Helpful?

  9. How do you remove the coverslip when using Fluoroshield (Product Nos. F6182, F5932, F6057)?

    1 answer
    1. The coverslip can be removed before sealing the edges. Soak slide in warm (37  °C) water for a few minutes. Carefully and slowly move the coverslip. Soak in water for an additional few minutes to remove coverslip. Rinse slide several times with warm water to remove all mounting medium. The slide can be remounted again.

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service