Merck
SAMHD1's protein expression profile in humans.
Journal of leukocyte biology (2015-02-04)
Sarah Schmidt, Kristina Schenkova, Tarek Adam, Elina Erikson, Judith Lehmann-Koch, Serkan Sertel, Bruno Verhasselt, Oliver T Fackler, Felix Lasitschka, Oliver T Keppler
ABSTRACT
The deoxynucleoside triphosphate triphosphohydrolase and 3' → 5' exonuclease SAMHD1 restricts HIV-1 infection in noncycling hematopoietic cells in vitro, and SAMHD1 mutations are associated with AGS. Little is known about the in vivo expression and functional regulation of this cellular factor. Here, we first assessed the SAMHD1 protein expression profile on a microarray of 25 human tissues from >210 donors and in purified primary cell populations. In vivo, SAMHD1 was expressed in the majority of nucleated cells of hematopoietic origin, including tissue-resident macrophages, DCs, pDCs, all developmental stages of thymic T cells, monocytes, NK cells, as well as at lower levels in B cells. Of note, SAMHD1 was abundantly expressed in HIV target cells residing in the anogenital mucosa, providing a basis for its evaluation as a cellular factor that may impact the efficiency of HIV transmission. Next, we examined the effect of the activation status and proinflammatory cytokine treatment of cells on expression and phosphorylation of SAMHD1. Activated, HIV-susceptible CD4(+) T cells carried pSAMHD1(T592), whereas resting CD4(+) T cells and macrophages expressed the unphosphorylated protein with HIV-restrictive activity. Surprisingly, stimulation of these primary cells with IFN-α, IFN-γ, IL-4, IL-6, IL-12, IL-18, IL-27, or TNF-α affected neither SAMHD1 expression levels nor threonine 592 phosphorylation. Only IL-1β moderately down-regulated SAMHD1 in activated CD4(+) T cells. Taken together, this study establishes the first cross-sectional protein expression profile of SAMHD1 in human tissues and provides insight into its cell cycle-dependent phosphorylation and unresponsiveness to multiple proinflammatory cytokines.
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Trimethylaluminum, 97%
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems