Merck
  • Silanization-Based Zeolite Crystallization: Participation Degree and Pathway.
Silanization-Based Zeolite Crystallization: Participation Degree and Pathway.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-16)
Yueer Yan, Arepati Azhati, Xiao Guo, Yahong Zhang, Yi Tang
ABSTRACT
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that organosilane plays the role of an inhibitor in the silanization-based zeolite preparation, and this inhibition effect was determined by its participation degree. An organosilane with a high participation degree can result in the prolongation of nucleation and growth periods of zeolite as well as the variation of product properties. More importantly, a dynamic participation pathway of organosilane is proposed, that is, the growth of zeolite is accompanied by the continuous removal of organosilane, leading to an increase of product crystallinity as well as the decrease of mesoporosity. This study gives a new insight into the role that organosilane plays in zeolite crystallization, which will help to direct the rational selection of organosilane and design of crystallization condition for the optimal synthesis of hierarchical zeolites.
Sigma-Aldrich
Triethoxymethylsilane, technical grade, 90%
Sigma-Aldrich
Tetrapropylammonium hydroxide solution, 1.0 M in H2O
Sigma-Aldrich
Triethoxy(octyl)silane, deposition grade, 97%, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Triethoxyphenylsilane, 98%
Sigma-Aldrich
Triethoxy(octyl)silane, 97%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Triethoxymethylsilane, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Diethoxydimethylsilane, 97%
Sigma-Aldrich
Isobutyltriethoxysilane, ≥95%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Aluminum isopropoxide, ≥98%
Sigma-Aldrich
3-Aminopropyl(diethoxy)methylsilane, 97%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Aluminum isopropoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Triethoxyvinylsilane, 97%
Sigma-Aldrich
Ethoxytrimethylsilane, 98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Autocamtide 2-related inhibitory peptide, ≥97% (HPLC), lyophilized powder
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Triethoxyvinylsilane, ≥98%, deposition grade