Skip to Content
Merck
All Photos(3)

Key Documents

220418

Sigma-Aldrich

Aluminum isopropoxide

≥98%

Synonym(s):

AIP, Aluminum isopropylate, Aluminum triisopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al[OCH(CH3)2]3
CAS Number:
Molecular Weight:
204.24
Beilstein:
3910275
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

form

solid

reaction suitability

core: aluminum
reagent type: catalyst

mp

128-133 °C (lit.)

density

1.035 g/mL at 25 °C (lit.)

SMILES string

CC(C)O[Al](OC(C)C)OC(C)C

InChI

1S/3C3H7O.Al/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

SMZOGRDCAXLAAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum isopropoxide is a versatile andinexpensive Lewis acid catalyst and dehydrating agent widely used in the fieldof organic transformations, sol-gel processing, and rechargeable batteries.

Application

Aluminum isopropoxide can be used:
  • As a precursor to synthesize alumina nanoparticles and multimetallic oxides via sol-gel processing.
  • As a catalyst for various organic transformations such as Meerwein–Ponndorf–Verley–Oppenauer (MPVO) reactions and ring-opening polymerization of lactides and lactones.
  • As coating material to fabricate electrodes for lithium-ion batteries toenhance cycling stability and performance.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiancong Liu et al.
Science advances, 3(5), e1603171-e1603171 (2017-06-01)
Thermally activated delayed fluorescence (TADF) materials are inspiring intensive research in optoelectronic applications. To date, most of the TADF materials are limited to metal-organic complexes and organic molecules with lifetimes of several microseconds/milliseconds that are sensitive to oxygen. We report
Activity and stability studies of H-transfer reduction reactions of aldehydes and ketones over aluminium isopropoxide heterogenised catalysts
Atika Muhammad, et al.
Royal Society of Chemistry Advances, 12, 33970-33980 (2022)
Enhanced cathode performance:Mixed Al2O3 and LiAlO2 coating of Li1.2Ni0.13Co0.13Mn0.54O2
Shengde Dong, et al.
ACS Applied Materials & Interfaces, 12, 38153-38162 (2020)
Mechanochemical Nonhydrolytic Sol?Gel-Strategy for the Production of Mesoporous Multimetallic Oxides
Zihao Zhang, et al.
Chemistry of Materials, 31, 5529-5536 (2019)
Zhehao Huang et al.
Nature communications, 11(1), 3762-3762 (2020-07-30)
Zeolites have unique pore structures of molecular dimensions and tunable compositions, making them ideal for shape selective catalysis and separation. However, targeted synthesis of zeolites with new pore structures and compositions remains a key challenge. Here, we propose an approach

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service