Merck
Self-Assembly of a Tripod Aromatic Rod into Stacked Planar Networks.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-07)
Ho-Joong Kim, Yongju Kim, Sung Cho, Myongsoo Lee
ABSTRACT
Threefold symmetric rigid-core molecules with an internally grafted poly(ethylene oxide) (PEO) chain were synthesized, and their self-assembled structures were characterized using differential scanning calorimetry, TEM, and 1D and 2D X-ray scatterings in the solid state. The tripod compounds based on short PEO chains (n=8, 13, 17, 21), self-assemble into 2D channel-like network structures, whereas the compound with the longest PEO chain (n=34) forms a lamellar liquid crystalline phase. The interiors of the channel structures are filled with flexible PEO chains along the double-walled aromatic circumference. In these channel-like networks, three aromatic rods connected in the meta-position to each other are superimposed in parallel to other adjacent molecules to form the double-walled aromatic frameworks stacked perpendicular to the resulting channels. These are novel examples of supramolecular channel-like structures developed using amphiphilic diblock molecules based on a threefold symmetric rigid scaffold.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Iodine monochloride solution, 1 M in acetic acid
Sigma-Aldrich
Boron tribromide, ReagentPlus®, 99.9%
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Boron tribromide solution, 1.0 M in heptane
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Boron tribromide solution, 1.0 M in methylene chloride
Sigma-Aldrich
Boron tribromide solution, 1.0 M in hexanes
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Boron tribromide, ReagentPlus®, ≥99%
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
Iodine monochloride, 99.998% trace metals basis
Sigma-Aldrich
4-Biphenylboronic acid, ≥95.0%
Sigma-Aldrich
Boron tribromide, ≥99.99%
Sigma-Aldrich
2,4,6-Tribromophenol, 99%