Merck
  • Amelioration of Bleomycin-induced Pulmonary Fibrosis of Rats by an Aldose Reductase Inhibitor, Epalrestat.
Amelioration of Bleomycin-induced Pulmonary Fibrosis of Rats by an Aldose Reductase Inhibitor, Epalrestat.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology (2015-09-04)
Xianwei Li, Yuanyuan Shen, Yining Lu, Jieren Yang
ABSTRACT
Aldose reductase (AR) is known to play a crucial role in the mediation of diabetic and cardiovascular complications. Recently, several studies have demonstrated that allergen-induced airway remodeling and ovalbumin-induced asthma is mediated by AR. Epalrestat is an aldose reductase inhibitor that is currently available for the treatment of diabetic neuropathy. Whether AR is involved in pathogenesis of pulmonary fibrosis and whether epalrestat attenuates pulmonary fibrosis remains unknown. Pulmonary fibrosis was induced by intratracheal instillation of bleomycin (5 mg/kg) in rats. Primary pulmonary fibroblasts were cultured to investigate the proliferation by BrdU incorporation method and flow cytometry. The expression of AR, TGF-β1, α-SMA and collagen I was analyzed by immunohistochemisty, real-time PCR or western blot. In vivo, epalrestat treatment significantly ameliorated the bleomycin-mediated histological fibrosis alterations and blocked collagen deposition concomitantly with reversing bleomycin-induced expression up-regulation of TGF-β1, AR, α-SMA and collagen I (both mRNA and protein). In vitro, epalrestat remarkably attenuated proliferation of pulmonary fibroblasts and expression of α-SMA and collagen I induced by TGF-β1, and this inhibitory effect of epalrestat was accompanied by inhibiting AR expression. Knockdown of AR gene expression reversed TGF-β1-induced proliferation of fibroblasts, up-regulation of α-SMA and collagen I expression. These findings suggest that AR plays an important role in bleomycin-induced pulmonary fibrosis, and epalrestat inhibited the progression of bleomycin-induced pulmonary fibrosis is mediated via inhibiting of AR expression.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b8
Sigma-Aldrich
MISSION® esiRNA, targeting human AREG
Sigma-Aldrich
Epalrestat, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human AKR1B1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Areg
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fdxr
Sigma-Aldrich
MISSION® esiRNA, targeting human AR
Sigma-Aldrich
MISSION® esiRNA, targeting human FDXR