Merck
  • "Dilute-and-shoot" rapid-separation liquid chromatography tandem mass spectrometry method for fast detection of thiodiglycolic acid in urine.
"Dilute-and-shoot" rapid-separation liquid chromatography tandem mass spectrometry method for fast detection of thiodiglycolic acid in urine.
European journal of mass spectrometry (Chichester, England) (2015-11-19)
Timur Baygildiev, Arcady Braun, Andrey Stavrianidi, Igor Rodin, Oleg Shpigun, Igor Rybalchenko, Irina Ananieva
ABSTRACT
A new sensitive rapid-separation liquid chromatography tandem mass spectrometry approach for the determination of thiodiglycolic acid (TDGA) in urine has been developed. The use of the "dilute-and-shoot" method helps to shorten the sample preparation stage and provides a sensitive and direct approach for TDGA determination in urine. Chromatographic separation of the analyte and other urine compounds was achieved using a reverse-phase liquid chromatography column with mobile phases consisting of 0.1% formic acid in water and acetonitrile in a gradient elution mode. For the identification and quantification of TDGA electrospray ionization-tandem mass spectrometry monitoring, two precursor-to-product ion transitions were used. The method demonstrates good linearity and has a detection limit of 50 ng mL⁻¹ in urine.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%