Merck
  • Deletion of Panx3 Prevents the Development of Surgically Induced Osteoarthritis.
Deletion of Panx3 Prevents the Development of Surgically Induced Osteoarthritis.
Journal of molecular medicine (Berlin, Germany) (2015-07-04)
Paxton M Moon, Silvia Penuela, Kevin Barr, Sami Khan, Christopher L Pin, Ian Welch, Mukundan Attur, Steven B Abramson, Dale W Laird, Frank Beier
ABSTRACT
Osteoarthritis (OA) is a highly prevalent, disabling joint disease with no existing therapies to slow or halt its progression. Cartilage degeneration hallmarks OA pathogenesis, and pannexin 3 (Panx3), a member of a novel family of channel proteins, is upregulated during this process. The function of Panx3 remains poorly understood, but we consistently observed a strong increase in Panx3 immunostaining in OA lesions in both mice and humans. Here, we developed and characterized the first global and conditional Panx3 knockout mice to investigate the role of Panx3 in OA. Interestingly, global Panx3 deletion produced no overt phenotype and had no obvious effect on early skeletal development. Mice lacking Panx3 specifically in the cartilage and global Panx3 knockout mice were markedly resistant to the development of OA following destabilization of medial meniscus surgery. These data indicate a specific catabolic role of Panx3 in articular cartilage and identify Panx3 as a potential therapeutic target for OA. Lastly, while Panx1 has been linked to over a dozen human pathologies, this is the first in vivo evidence for a role of Panx3 in disease. Panx3 is localized to cartilage lesions in mice and humans. Global Panx3 deletion does not result in any developmental abnormalities. Mice lacking Panx3 are resistant to the development of osteoarthritis. Panx3 is a novel therapeutic target for the treatment of osteoarthritis.
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the Biological Stain Commission, Dye content 85 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Methyl Green, zinc chloride salt, for microscopy (Bact., Bot., Hist.)