Merck
Prognostic relevance of LGALS3BP in human colorectal carcinoma.
Journal of translational medicine (2015-07-30)
Enza Piccolo, Nicola Tinari, Domenica D'Addario, Cosmo Rossi, Valentina Iacobelli, Rossana La Sorda, Rossano Lattanzio, Maurizia D'Egidio, Annalisa Di Risio, Mauro Piantelli, Pier Giorgio Natali, Stefano Iacobelli
ABSTRACT
A previous report has shown that LGALS3BP (also known as 90K or Mac-2 BP) has antitumor activity in colorectal cancer (CRC) via suppression of Wnt signalling with a novel mechanism of ISGylation-dependent ubiquitination of β-catenin. The role of LGALS3BP in CRC prognosis was investigated. The role of LGALS3BP on CRC progression and clinical prognosis was analyzed by combining cell cultures, in vitro assays, and immunohistochemistry. Silencing of LGALS3BP in HCT-116 human colon cancer cells resulted in enhanced β-catenin expression that was reversed by addition of human recombinant LGALS3BP. Moreover, intra-tumor delivery of LGALS3BP reduced tumor growth of xenografts originating from LGALS3BP-silenced HCT-116 cells. Finally, in a series of 196 CRC patients, LGALS3BP expression in tumor tissue associated with clinical outcome. Patients with high LGALS3BP expression had lower risk of relapse and a longer overall survival time than those with low LGALS3BP expression. Multivariate analyses confirmed LGALS3BP expression status as the only independent prognostic factor of survival. These results provide evidence that low expression of LGALS3BP participates in malignant progression of CRC and implicates poor prognosis, highlighting its augmentation as a potential therapeutic approach.
Sigma-Aldrich
Cyanogen bromide, 99.995% trace metals basis
Sigma-Aldrich
Cyanogen bromide, reagent grade, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine