Merck
  • Lack of ADAM2, CALR3 and SAGE1 Cancer/Testis Antigen Expression in Lung and Breast Cancer.
Lack of ADAM2, CALR3 and SAGE1 Cancer/Testis Antigen Expression in Lung and Breast Cancer.
PloS one (2015-08-08)
Emeaga Maheswaran, Christina B Pedersen, Henrik J Ditzel, Morten F Gjerstorff
ABSTRACT
Immunotherapy is emerging as a supplement to conventional cancer treatment, and identifying antigen targets for specific types of cancer is critical to optimizing therapeutic efficacy. Cancer/testis antigens are highly promising targets for immunotherapy due to their cancer-specific expression and antigenic properties, but the expression patterns of most of the more than 200 identified cancer/testis antigens in various cancers remain largely uncharacterized. In this study, we investigated the expression of the cancer/testis antigens ADAM2, CALR3 and SAGE1 in lung and breast cancer, the two most frequent human cancers, with the purpose of providing novel therapeutic targets for these diseases. We used a set of previously uncharacterized antibodies against the cancer/testis antigens ADAM2, CALR3 and SAGE1 to investigate their expression in a large panel of normal tissues as well as breast and lung cancers. Staining for the well-characterized MAGE-A proteins was included for comparison. Immunohistochemical staining confirmed previous mRNA analysis demonstrating that ADAM2, CALR3 and SAGE1 proteins are confined to testis in normal individuals. Negative tissues included plancenta, which express many other CT antigens, such as MAGE-A proteins. Surprisingly, we detected no ADAM2, CALR3 and SAGE1 in the 67 lung cancers (mainly non-small lung cancer) and 189 breast cancers, while MAGE-A proteins were present in 15% and 7-16% of these tumor types, respectively. Treatment with DNA methyltransferase inhibitors has been proposed as an attractive strategy to increase the expression of cancer/testis antigens in tumors before immunotargeting; however, neither ADAM2, CALR3 nor SAGE1 could be significantly induced in lung and breast cancer cell lines using this strategy. Our results suggest that ADAM2, CALR3 and SAGE1 cancer/testis antigens are not promising targets for immunotherapy of breast and lung cancer.
Sigma-Aldrich
Anti-SAGE1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-ADAM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission